DiluFlux™ Pro Automated Gravimetric Dilutor
Actual product may vary
DiluFlux™ Pro Automated Gravimetric Dilutor
DiluFlux™ Pro Automated Gravimetric Dilutor
Thermo Scientific™

DiluFlux™ Pro Automated Gravimetric Dilutor

Accurately dispense media into suitable containers according to a set dilution using this gravimetric diluter which allows the use of two diluents.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberIncludesModel
DB4100ASimple Dispensing ArmDiluFlux Pro-3
DB4150ARobotic Dispensing ArmDiluFlux Pro-5
Catalog number DB4100A
Price (USD)
-
Includes:
Simple Dispensing Arm
Model:
DiluFlux Pro-3
Accurately dispense broth medium into suitable containers for food testing using the Thermo Scientific™ DiluFlux™ Pro Automated Gravimetric Dilutor. DiluFlux Pro Dilutors are compact, easy-to-use, and designed to deliver precise quantities of diluents rapidly with the touch of a button, so that you can work more confidently and accurately. When combined with the Thermo Scientific™ Peristaltic Pump and Thermo Scientific™ Homogenizer, your lab bench will be equipped with a flexible automated culture media solution to increase your productivity.

Automate the dilution step of your Thermo Scientific™ Dry-Bags™ Enrichment Media, as well as other formats, for maximum efficiency. The easy-to-use DiluFlux Pro Gravimetric Dilutors allow the simultaneous use of two diluents by having dual dispensing heads that are fed by dual peristaltic pumps mounted into the unit.

  • Intuitive: simply press two keys, tare and go, and the DiluFlux Pro Dilutor will automatically dilute your sample to the desired dilution factor
  • Accurate: featuring Watson-Marlow™ peristaltic pumps, high-tech flow regulators, and top of the line weighing platforms for trusted reproducibility and repeatability
  • Versatile: compatible with many prepared media formats, the easy-to clean, stainless steel DiluFlux Pro Dilutor is an ideal fit for any food testing laboratory
  • High-quality: the DiluFlux Pro Dilutors are ISO 7218, ISO 6887-1, FDA BAM-compliant and are made in France under ISO 9001:2008 certification

Not all products are available for sale in all territories. Please inquire.

For testing of Food and Environmental samples only.
For testing of Food and Environmental samples only.
Specifications
DescriptionDiluFlux Pro-3 Automated Gravimetric Dilutor w/ simple (non-robotic) dispensing arm
Dimensions (L x W x H)41.5 x 31 x 40 cm
Electrical Requirements100/127 V 220/240 V 50/60 Hz
For Use With (Equipment)Dry-Bags
Frequency50/60 Hz
IncludesSimple Dispensing Arm
MaterialStainless Steel (Case)
ModelDiluFlux Pro-3
Overall Height (Metric)40 cm
Overall Length (Metric)41.5 cm
Overall Width (Metric)31 cm
Shipping Weight (Metric)10.0 kg
Voltage100/240 V
Unit SizeEach