Confidently perform susceptibility testing using one of the latest antimicrobials with the Thermo Scientific™ Sensititre™ Delafloxacin DELAXN AST Plate. Address your formulary needs with this micro-broth dilution plate for antimicrobial susceptibility testing that meets CLSI standards.
Obtain true minimum inhibitory concentration (MIC) results to both inform targeted therapy decisions for resistant infections with otherwise limited or no treatment options, and to support antimicrobial stewardship to preserve effectiveness.
0.5 McFarland Standard | H2O (Part No. T3339) |
Antimicrobial Agents | Delafloxacin |
AST Type | MIC |
Broth Type | MHB (Part No. T3462) |
Concentration | Multiple |
Description | Sensititre™ Delafloxacin DELAXN Plate |
Format | Multi-antibiotic Microtitre Panels |
Product Type | Delafloxacin AST Plate |
Test Type | Delafloxacin Susceptibility, MIC |
Incubation Conditions | 35°C O2 |
Incubation Time | Refer to Product Insert |
Isolates Per Plate | 6 |
Packaging Type | 10 x microtitre panels |
Quantity | 10/Pk. |
Reading Options | Manual |
Transfer Volume | Refer to Product Insert |
Volume Per Well | 50 μL |
Unit Size | Each |
Catalog Number | Specifications | Unit Size | Price (USD) | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|
DELAXN | Each | Request A Quote | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
|
Expand treatment options for acute bacterial skin and skin structure infections (ABSSSI) by using the Sensititre Delafloxacin DELAXN AST Plate for accurate, economical delafloxacin susceptibility results.
The Sensititre System is a leader in AST solutions, offering flexible, customizable testing options for laboratories of all sizes.
Not all products are available for sale in all territories. Please inquire.
VersaTREK™ and Sensititre™ products are now part of the Thermo Scientific brand