NERL™ Diluent 2 Hematology Reagent for Flow Cytometry
Thermo Scientific™

NERL™ Diluent 2 Hematology Reagent for Flow Cytometry

Hematology reagents for use with various Beckman Coulter& analyzers. Multi-Purpose Blood diluent.
Have Questions?
Catalog number DIL5522
Price (CLP)
-
 
Multi-Purpose Blood Diluent for use on Coulter Analyzers. Cut costs by replacing manufacturer's reagents with Thermo Scientific™ NERL™ Diluent 2 Hematology Reagent for Flow Cytometry. This anti-microbial additive keeps reagent purity constant, with no added detergents.
For In Vitro Diagnostic Use.  
Specifications
DescriptionThermo Scientific NERL Diluent 2 Hematology Reagent
For Use With (Equipment)Coulter Analyzers not Performing WBC Differentials
Quantity20 L Cubitainer
Shelf Life18 months
TypeHematology Reagent
Unit SizeEach