Savant™ DNA SpeedVac™ Concentrator Kit, DNA120, 115V, 60Hz
Save time and money with Thermo Scientific™ Savant™ DNA SpeedVac™ Concentrators Kits. Everything you need for quick drying and concentrating of small sample volumes.
Have Questions?
Catalog number DNA120-115
Price (CLP)
-
Save time and money with compact Thermo Scientific™ Savant™ DNA SpeedVac™ Concentrators Kits. These complete systems provide everything you need to quickly dry and concentrate small sample volumes in one fully integrated package, sold under one part number.
Operating Features
Consistent performance: designed to ensure consistent drying times and reproducible results
Simplicity: single program supports all DNA/RNA applications
User-friendly interface: with temperature/time settings
Three drying rates: ambient, ∽43°C, ∽65°C
Dual digital timers: independent control of heat/run times
Sample Protection
Dry at ambient temperatures or at very low heat to avoid sample denaturing
Oil-free diaphragm pump provides gentle transition from atmosphere to ultimate vacuum setting with no bumping
Safety Features
PTFE-coated chamber resists chemicals and cleans easily
Latched lid ensures user safety during operation
Unit and pump are virtually maintenance-free
Optional chemical traps and cartridges trap radioactivity and ammonia
Solvents and Solvent Combinations:
DNA120
PCR buffers (aqueous)
Water
Ethanol and water
DNA-120-OP
Ammonium hydroxide
Isopropanel
PCR buffers (aqueous)
ISS-110
PCR buffers (aqueous)
Ethanol and water
Isopropanol
DNA 120 SpeedVac Kit
Corrosion-resistant, PTFE coated chamber provides years of lasting service