QuickBag™ Tryptone Soya Broth (TSB)
QuickBag™ Tryptone Soya Broth (TSB)
QuickBag™ Tryptone Soya Broth (TSB)
QuickBag™ Tryptone Soya Broth (TSB)
Thermo Scientific™

QuickBag™ Tryptone Soya Broth (TSB)

Shorten your workflow with easy-to-prepare 2.7L or 4.5L media bags: simply merge the segments to reconstitute in two minutes.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
DQ0129A3 x 2.7 L
DQ0129B2 x 4.5 L
Catalog number DQ0129A
Price (USD)
-
Quantity:
3 x 2.7 L
Save time in preparing a highly nutritious versatile medium that supports fastidious organism growth using Thermo Scientific™ QuickBag™ Tryptone Soya Broth (TSB). The irradiated medium is supplied in transparent plastic bags—simply pop the seal to mix the water and medium segments to reconstitute in two minutes. QuickBag Enrichment Media are available in 2.7L and 4.5L sizes to suit your workflow. Medium can be dispensed through the outlet tube as required. Trytone Soya Broth conforms to formulations detailed in international pharmacopoeias.1, 2,3,4

Shorten your workflow with QuickBag UVM1 Broth. QuickBag Enrichment Media is quick, easy-to-prepare, and highly adaptable to laboratory workflows and automated equipment, such as dilutors. No further supplementation is required.

Cost-effective: no need for weighing of media or sterilization, reducing the burden of media preparation, which frees up resources and equipment.

Flexible: segmented structure allows advanced preparation by filling one segment with deionized water from which QuickBag Enrichment Media can be reconstituted within two minutes. Pre-sterilized water may also be added under aseptic conditions.

Easy-to-use: fill and dispense with minimum training.

Energy saving:
gamma irradiated, eliminating the need for costly thermal processing.

Safe: eliminate handling of glassware, weighing of culture media, and the need to add supplementary reagents. Empty bags can be disposed of with normal laboratory waste.

Reduces waste: unprepared QuickBag Enrichment Media can be kept for up to 24 months. Rehydrated media has a shelf life of 10 days.

Quality guaranteed: compliant with ISO 11133 and ISO 7218. Quality certificates are available for every batch, reducing the QC testing needed.

Space-saving:
conveniently packed in flat boxes, QuickBag Enrichment Media takes up relatively little space compared to prepared media and can be made up as required.

For Laboratory Use
For Laboratory Use

General References:

  1. British Pharmacopoeia (2004).
  2. Japanese Pharmacopoeia. 15th Edition. (2006).
  3. US Pharmacopoeia 30 (2008).
  4. European Pharmacopoeia. 6.1 (2008).
Specifications
DescriptionQuickBag Tryptone Soya Broth (TSB)
FormatConvenience Format
Media TypeTryptone Soya Broth
Quantity3 x 2.7 L
Shelf Life2 years (rehydrated 10 Days)
Product TypeNutrient Broth
Unit SizeEach
Contents & Storage
10°C to 30°C