QuickBag™ UVM1 Broth
QuickBag™ UVM1 Broth
QuickBag™ UVM1 Broth
QuickBag™ UVM1 Broth
Thermo Scientific™

QuickBag™ UVM1 Broth

Shorten your workflow with easy-to-prepare 2.7L or 4.5L media bags: simply merge the segments to reconstitute in two minutes.
Have Questions?
보기 방식 변경buttonViewtableView
카탈로그 번호수량
DQ0863B2 x 4.5 L
DQ0863A3 x 2.7 L
카탈로그 번호 DQ0863B
제품 가격(KRW)
-
수량:
2 x 4.5 L
Save time in preparing selective enrichment medium for Listeria monocytogenes, while conforming to the USDA-FSIS method, using Thermo Scientific™ QuickBag™ UVM1 Broth. The irradiated medium is supplied in transparent plastic bags—simply pop the seal to mix the water and medium segments to reconstitute in two minutes. QuickBag Enrichment Media are available in 2.7L and 4.5L sizes to suit your workflow. Medium can be dispensed through the outlet tube as required.

Shorten your workflow with QuickBag UVM1 Broth. QuickBag Enrichment Media is quick, easy-to-prepare, and highly adaptable to laboratory workflows and automated equipment, such as dilutors. No further supplementation is required.

Cost-effective: no need for weighing of media or sterilization, reducing the burden of media preparation, which frees up resources and equipment.

Flexible: segmented structure allows advanced preparation by filling one segment with deionized water from which QuickBag Enrichment Media can be reconstituted within two minutes. Pre-sterilized water may also be added under aseptic conditions.

Easy-to-use: fill and dispense with minimum training.

Energy saving:
gamma irradiated, eliminating the need for costly thermal processing.

Safe: eliminate handling of glassware, weighing of culture media, and the need to add supplementary reagents. Empty bags can be disposed of with normal laboratory waste.

Reduces waste: unprepared QuickBag Enrichment Media can be kept for up to 24 months. Rehydrated media has a shelf life of 10 days.

Quality guaranteed: compliant with ISO 11133 and ISO 7218. Quality certificates are available for every batch, reducing the QC testing needed.

Space-saving:
conveniently packed in flat boxes, QuickBag Enrichment Media takes up relatively little space compared to prepared media and can be made up as required.

실험실 전용.
실험실 전용.
사양
설명QuickBag UVM1 Broth
형식Convenience Format
미디어 유형UVM1 Broth
수량2 x 4.5 L
유통 기한2 years (rehydrated 10 Days)
제품 유형Nutrient Broth
Unit SizeEach
구성 및 보관
10°C to 30°C