Thermo Scientific™

Streptococcal Grouping Kit using Latex Agglutination

Catalog number: DR0587G
Thermo Scientific™

Streptococcal Grouping Kit using Latex Agglutination

Catalog number: DR0587G

Rapidly identify streptococcal groups A, B, C, D, F, and G by latex agglutination using Thermo Scientific™ Oxoid™ Streptococcal Grouping Kit. This rapid latex agglutination test is designed to deliver simple, rapid and sensitive results for the identification of streptococcal groups A, B, C, D, F, and G . The test utilizes latex particles coated with group-specific antibodies which will agglutinate in the presence of homologous antigens.

 
Catalog Number
DR0587G
Unit Size
Each
Description
Streptococcal Grouping Reagent B
Price (USD)
Full specifications
DescriptionStreptococcal Grouping Reagent B
TypeReagents for Streptococcus Testing
Organism GroupGroup A, B, C, D, F and G
Packaging Type50 Tests/Vial
Quantity50 Tests
Unit SizeEach
Catalog Number
Unit Size
Each
Description
Streptococcal Grouping Kit
Price (USD)
Full specifications
DescriptionStreptococcal Grouping Kit
Detectable AnalytesStreptococcal groups A, B, C, D, F, and G
TypeStreptococcus Test Kit
Organism GroupLancefield A, B, C, D, F and G
Quantity50 Tests/Kit
Unit SizeEach
Catalog Number
Unit Size
Each
Description
Streptococcal Grouping Reagent A
Price (USD)
Full specifications
DescriptionStreptococcal Grouping Reagent A
TypeReagents for Streptococcus Testing
Organism GroupGroup A, B, C, D, F and G
Packaging Type50 Tests/Vial
Quantity50 Tests
Unit SizeEach
Catalog Number
Unit Size
Each
Description
Streptococcal Grouping Reagent C
Price (USD)
Full specifications
DescriptionStreptococcal Grouping Reagent C
TypeReagents for Streptococcus Testing
Organism GroupGroup A, B, C, D, F and G
Packaging Type50 Tests/Vial
Quantity50 Tests
Unit SizeEach
Catalog Number
Unit Size
Each
Description
Streptococcal Grouping Reagent D
Price (USD)
Full specifications
DescriptionStreptococcal Grouping Reagent D
TypeReagents for Streptococcus Testing
Organism GroupGroup A, B, C, D, F and G
Packaging Type50 Tests/Vial
Quantity50 Tests
Unit SizeEach
Catalog Number
Unit Size
Each
Description
Streptococcal Grouping Reagent F
Price (USD)
Full specifications
DescriptionStreptococcal Grouping Reagent F
TypeReagents for Streptococcus Testing
Organism GroupGroup A, B, C, D, F and G
Packaging Type50 Tests/Vial
Quantity50 Tests
Unit SizeEach
Catalog Number
Unit Size
Each
Description
Streptococcal Grouping Reagent G
Price (USD)
Full specifications
DescriptionStreptococcal Grouping Reagent G
TypeReagents for Streptococcus Testing
Organism GroupGroup A, B, C, D, F and G
Packaging Type50 Tests/Vial
Unit SizeEach
Catalog Number
Unit Size
Each
Description
Polyvalent Positive Control; 50 tests
Price (USD)
Full specifications
Control SetsPositive
DescriptionPolyvalent Positive Control; 50 tests
TypePositive Control
MaterialPolyvalent
Organism GroupGroup A, B, C, D, F and G
Quantity50 Tests
Unit SizeEach
Catalog Number
Unit Size
Each
Description
Streptococcal Grouping Enzyme Extraction Reagent
Price (USD)
Full specifications
DescriptionStreptococcal Grouping Enzyme Extraction Reagent
TypeReagents for Streptococcus Testing
For Use With (Application)Enzyme Immunoassay Detection
Organism GroupGroup A, B, C, D, F and G
Packaging Type2 Vials
Quantity2 x 50 Tests
Unit SizeEach
Showing 9 of 9
Catalog NumberSpecificationsUnit SizeDescriptionPrice (USD)
DR0587GFull specifications
EachStreptococcal Grouping Reagent BRequest A Quote
DescriptionStreptococcal Grouping Reagent B
TypeReagents for Streptococcus Testing
Organism GroupGroup A, B, C, D, F and G
Packaging Type50 Tests/Vial
Quantity50 Tests
Unit SizeEach
DR0585AFull specifications
EachStreptococcal Grouping KitRequest A Quote
DescriptionStreptococcal Grouping Kit
Detectable AnalytesStreptococcal groups A, B, C, D, F, and G
TypeStreptococcus Test Kit
Organism GroupLancefield A, B, C, D, F and G
Quantity50 Tests/Kit
Unit SizeEach
DR0586GFull specifications
EachStreptococcal Grouping Reagent ARequest A Quote
DescriptionStreptococcal Grouping Reagent A
TypeReagents for Streptococcus Testing
Organism GroupGroup A, B, C, D, F and G
Packaging Type50 Tests/Vial
Quantity50 Tests
Unit SizeEach
DR0588GFull specifications
EachStreptococcal Grouping Reagent CRequest A Quote
DescriptionStreptococcal Grouping Reagent C
TypeReagents for Streptococcus Testing
Organism GroupGroup A, B, C, D, F and G
Packaging Type50 Tests/Vial
Quantity50 Tests
Unit SizeEach
DR0589GFull specifications
EachStreptococcal Grouping Reagent DRequest A Quote
DescriptionStreptococcal Grouping Reagent D
TypeReagents for Streptococcus Testing
Organism GroupGroup A, B, C, D, F and G
Packaging Type50 Tests/Vial
Quantity50 Tests
Unit SizeEach
DR0590GFull specifications
EachStreptococcal Grouping Reagent FRequest A Quote
DescriptionStreptococcal Grouping Reagent F
TypeReagents for Streptococcus Testing
Organism GroupGroup A, B, C, D, F and G
Packaging Type50 Tests/Vial
Quantity50 Tests
Unit SizeEach
DR0591GFull specifications
EachStreptococcal Grouping Reagent GRequest A Quote
DescriptionStreptococcal Grouping Reagent G
TypeReagents for Streptococcus Testing
Organism GroupGroup A, B, C, D, F and G
Packaging Type50 Tests/Vial
Unit SizeEach
DR0592GFull specifications
EachPolyvalent Positive Control; 50 testsRequest A Quote
Control SetsPositive
DescriptionPolyvalent Positive Control; 50 tests
TypePositive Control
MaterialPolyvalent
Organism GroupGroup A, B, C, D, F and G
Quantity50 Tests
Unit SizeEach
DR0593GFull specifications
EachStreptococcal Grouping Enzyme Extraction ReagentRequest A Quote
DescriptionStreptococcal Grouping Enzyme Extraction Reagent
TypeReagents for Streptococcus Testing
For Use With (Application)Enzyme Immunoassay Detection
Organism GroupGroup A, B, C, D, F and G
Packaging Type2 Vials
Quantity2 x 50 Tests
Unit SizeEach
Showing 9 of 9

Streptococci are facultatively anaerobic, catalase-negative, Gram-positive cocci that occur as pairs or chains. They cause a wide variety of clinical diseases including pharyngitis, scarlet fever, otitis media, impetigo, meningitis, neonatal sepis, pneumonia, endocarditis, osteomyelitis, and necrotizing fasciitis.

Use this rapid, simple and sensitive test for the identification of streptococcal groups A, B, C, D, F, and G.

  • Rapid—results in two minutes
  • Simple—easy to implement
  • Visual—easy-to-read results
  • Sensitive—use of latex technology makes this test more sensitive
  • All reagents available separately

Lancefield showed that the majority of pathogenic streptococci possess specific carbohydrate antigens, which permit the classification of streptococci into groups. These streptococcal group antigens can be extracted from the cells and their presence demonstrated with latex particles previously coated with group-specific antibodies. These latex particles will agglutinate in the presence of homologous antigen, but will remain in smooth suspension in the absence of such antigen. The Oxoid Streptococcal Grouping Kit is such a latex agglutination test for the identification of the streptococcal groups and reagents are provided for groups A, B, C, D, F and G. The use of a enzymatic extraction procedure considerably shortens the time required for antigen extraction and much improves the antigen yield, partially for Group D streptococci.

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.

Figures