Staphylase™ Test
Thermo Scientific™
Staphylase™ Test
Rapidly identify Staphylococcus aureus with this slide agglutination test which detects the presence of the clumping factor from selective or nonselective culture media.
Have Questions?
Número do catálogoQuantity
DR0595A100 Tests/Kit
Número do catálogo DR0595A
Preço (BRL)
-
Quantity:
100 Tests/Kit
The generally accepted identifying characteristic of Staphylococcus aureus is the ability to produce free and bound coagulase (or clumping factor). The presence of clumping factor may be detected in a number of ways. The Oxoid Staphylase Test detects the presence of clumping factor through clumping of fibrinogen- sensitized sheep red blood cells1,2. The specificity of the reaction is ensured by a simultaneous test with a control reagent (unsensitized sheep red blood cells), when of course no clumping reaction should be observed.
Use this rapid agglutination test for the identification of Staphylococcus aureus by detection of clumping factor. Clumping factor is detected through agglutination of fibrinogen-sensitized sheep red blood cells. The specificity of the reaction is ensured by a simultaneous test with a control reagent (unsensitized sheep red blood cells) when no clumping reaction should be observed. A positive result is obtained if clumping of the test cell suspensions occurs during mixing. This indicates the presence of Staphylococcus aureus.
Included in the kit is a test reagent consisting of rabbit fibrogen-sensitized sheep red blood cells, control reagent consisting of unsensitized sheep red blood cells, and disposable reaction cards.
Each kit contains sufficient reagent for 100 tests.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.
Especificações
Control SetsPositive
DescriptionStaphylase Test
IncludesTest reagent, control reagent, disposable reaction cards. Each kit contains sufficient reagent for 100 tests.
Quantity100 Tests/Kit
Target Organism ClassStaphylococcus aureus
TypeStaphylococcus aureus Test Kit
Detectable AnalytesS. aureus
Unit SizeEach
Conteúdo e armazenamento
2°C to 8°C