Product Image
Thermo Scientific™
Oxoid™ Listeria Test Kit
Thermo Scientific™ Oxoid Listeria Kit is a rapid latex agglutination test used for presumptive identification of Listeria spp. in cultures.
Have Questions?
Catalog NumberQuantity
DR1126A50 Tests/Kit
Catalog number DR1126A
Price (USD)
-
Quantity:
50 Tests/Kit
Thermo Scientific™ Oxoid Listeria Kit is a rapid latex agglutination test used for presumptive identification of Listeria spp. in cultures.
Microtiter plate is inoculated with enriched pre-tested food samples and washed with monoclonal antibody conjugate which binds to Salmonella antigens forming an immune complex. 8 breakaway strips.
Specifications
DescriptionListeria Test Kit
Includes50 Tests/Kit
Quantity50 Tests/Kit
Sample TypePre-Tested Food
Target Organism ClassListeria Spp.
TypeListeria Test Kit
Detectable AnalytesListeria species
Unit SizeEach