Oxoid™ Dip-Slides Clinical
Oxoid™ Dip-Slides Clinical
Actual product may vary
Thermo Scientific™
Oxoid™ Dip-Slides Clinical
Make colony counting for the bacterial assessment of urine simple with the easily disposable Thermo Scientific™ Oxoid™ Dip-Slides Clinical.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantityFormatMedia TypeManufacturing Origin
DS0103A10/Pk.Dip SlideMacConkey Agar and CLED MediumUnited Kingdom
DS0102B100/Pk.Dip SlideMacConkey Agar and CLED MediumUnited Kingdom
GFD02A10/Pk.Dip SlideMacConkey /CLED/ Malt Extr+CAMItaly
Catalog number DS0103A
Price (TWD)
-
Request A Quote
Quantity:
10/Pk.
Format:
Dip Slide
Media Type:
MacConkey Agar and CLED Medium
Manufacturing Origin:
United Kingdom
Make colony counting for the bacterial assessment of urine simple with the easily disposable Thermo Scientific™ Oxoid™ Dip-Slides™ Clinical. Simple and safe for an unskilled operator to use, these dip slides are coated on both sides with a solid culture medium with the opportunity for different media to be carried on each side, making the process time- and cost-effective. This sterile dip slide can be inoculated in the ward or doctor's surgery and then returned to its plastic container for easy transport back to the laboratory.
Previous methods of detecting urinary tract infections depended on freshly voided urine being delivered promptly to a bacteriological laboratory for examination, either immediately or after a short period of refrigerated storage. The dip slide was developed as a logical extension of this method, using a microscope slide coated on each side with a different culture medium. With these dip slides, larger areas of media can be used for bacterial assessment of urine, making them time- and cost-effective.
  • Slide is made of easily disposable plastic with raised edges to ensure an even thickness of culture medium
  • Molded grid allows for easier colony counting
  • Slide is shaped to ensure drainage of fluid from the slide
  • Cap forms a convenient handle by which it may be held without risk of touching the culture medium
  • Bacteria chart is included, make examining growth simple for the operator and where laboratory facilities are limited
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products with a comprehensive line of media and diagnostic products, providing a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.
Specifications
FormatDip Slide
Media TypeMacConkey Agar and CLED Medium
Quantity10/Pk.
Manufacturing OriginUnited Kingdom
Product TypeSample Collection
Unit SizeEach
Contents & Storage
15°C to 25°C