Search
| Catalog Number | Format | Media Type | Manufacturing Origin |
|---|---|---|---|
| DS0167A | Dip Slide | MacConkey Agar # 3 + germicide | United Kingdom |
| DS0166A | Dip Slide | MacConkey Agar # 3 | United Kingdom |
| DS0170A | Dip Slide | OGYE Agar | United Kingdom |
| DS0168A | Dip Slide | VRGB Agar | United Kingdom |
| DS0169A | Dip Slide | VRBG Agar + germicide | United Kingdom |
| DS0147A | Dip Slide | TTC (Red Spot) | United Kingdom |
| DS0155A | Dip Slide | TTC (Red Spot)/Malt Ext Agar | United Kingdom |
Originally designed to determine bacterial counts on urine specimens, dip slides have been modified over the years by making the device double-sided, with one side a medium for Gram positive species and the other side a medium for Gram negatives. With these dip slides, the process for colony counting is time- and cost-effective, allowing exposure of the specimens to two different media simultaneously.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products with a comprehensive line of media and diagnostic products, providing a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.