Analytical Membrane Bands
Analytical Membrane Bands
Analytical Membrane Bands
Analytical Membrane Bands
Thermo Scientific™
Analytical Membrane Bands
Thermo Scientific Analytical Membrane Bands are perfect for both microbial QA and QC applications where membrane dispensers are used. Product is compatible with most common dispensers on the market, giving additional flexibility and security of supply from the leading lab and life sciences supplier.
Have Questions?
Cambiar vistabuttonViewtableView
Número de catálogoColorTamaño de poro
DS0600-0202Negro0,2 μm
DS0600-0102Blanco0,2 μm
DS0600-0108Blanco0,8 μm
DS0600-0145Blanco0,45 μm
DS0600-0208Negro0,8 μm
DS0600-0245Negro0,45 μm
Número de catálogo DS0600-0202
Precio (CLP)
-
Color:
Negro
Tamaño de poro:
0,2 μm
Thermo Scientific™ Analytical Membrane Bands are perfect for microbial QA/QC applications where membrane dispensers are used. Compatible with most common dispensers on the market, including MilliporeSigma™ and Sartorius™, giving added flexibility and security of supply. Boxes feature two marked orientations and tear-away perforations with a thumb-tab for quick and easy loading. Bands are numbered to indicate how many membranes remain to be dispensed. Membranes are gridded to improve accuracy and save time. Product has been validated to the highest standards.
Thermo Scientific™ Analytical Membrane Bands are perfect for both microbial quality assurance (QA) and quality control (QC) applications where membrane dispensers are used.
  • Product is compatible with most common dispenser platforms on the market, including MilliporeSigma™ and Sartorius™, giving additional flexibility and security of supply from the leading lab and life sciences supplier.
  • 47mm disks. Mixed cellulose esters (MCE). Available in both white and Black.
  • Each case of pre-sterilized Analytical Membrane Bands contains four boxes of 150 membranes per pack (600/case total).
  • Boxes feature two marked orientations and tear-away perforations with a thumb-tab for quick and easy loading.
  • Bands are numbered to indicate how many membranes remain to be dispensed.
  • Membranes are gridded to improve accuracy and save time.
  • Product has been validated to the highest standards, including ISO 7704 and ASTM F838.
Especificaciones
ColorNegro
Diámetro del disco (métrico)47 mm
Cantidad600/Cs
Material (membrana)MCE
Tamaño de poro0,2 μm
EsterilidadEstéril
Unit SizeCase of 600