Analytical Membrane Bands
Analytical Membrane Bands
Analytical Membrane Bands
Analytical Membrane Bands
Thermo Scientific™
Analytical Membrane Bands
Thermo Scientific Analytical Membrane Bands are perfect for both microbial QA and QC applications where membrane dispensers are used. Product is compatible with most common dispensers on the market, giving additional flexibility and security of supply from the leading lab and life sciences supplier.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberColorPore Size
DS0600-0208Black0.8 μm
DS0600-0102White0.2 μm
DS0600-0108White0.8 μm
DS0600-0145White0.45 μm
DS0600-0202Black0.2 μm
DS0600-0245Black0.45 μm
Catalog number DS0600-0208
Price (USD)
229.93
Online Exclusive
326.00
Save 96.07 (29%)
Case of 600
Color:
Black
Pore Size:
0.8 μm
Price (USD)
229.93
Online Exclusive
326.00
Save 96.07 (29%)
Case of 600
Thermo Scientific™ Analytical Membrane Bands are perfect for microbial QA/QC applications where membrane dispensers are used. Compatible with most common dispensers on the market, including MilliporeSigma™ and Sartorius™, giving added flexibility and security of supply. Boxes feature two marked orientations and tear-away perforations with a thumb-tab for quick and easy loading. Bands are numbered to indicate how many membranes remain to be dispensed. Membranes are gridded to improve accuracy and save time. Product has been validated to the highest standards.
Thermo Scientific™ Analytical Membrane Bands are perfect for both microbial quality assurance (QA) and quality control (QC) applications where membrane dispensers are used.
  • Product is compatible with most common dispenser platforms on the market, including MilliporeSigma™ and Sartorius™, giving additional flexibility and security of supply from the leading lab and life sciences supplier.
  • 47mm disks. Mixed cellulose esters (MCE). Available in both white and Black.
  • Each case of pre-sterilized Analytical Membrane Bands contains four boxes of 150 membranes per pack (600/case total).
  • Boxes feature two marked orientations and tear-away perforations with a thumb-tab for quick and easy loading.
  • Bands are numbered to indicate how many membranes remain to be dispensed.
  • Membranes are gridded to improve accuracy and save time.
  • Product has been validated to the highest standards, including ISO 7704 and ASTM F838.
Specifications
ColorBlack
Diameter (Metric) Disc47 mm
Quantity600/Cs.
Material (Membrane)MCE
Pore Size0.8 μm
SterilitySterile
Unit SizeCase of 600