Nalgene™ Polycarbonate, Validation Bottle
Thermo Scientific™

Nalgene™ Polycarbonate, Validation Bottle

For Use With: Perform validation testing; Autoclavable; Closure Material: Polypropylene; Color: White (Translucent)
Have Questions?
보기 방식 변경buttonViewtableView
카탈로그 번호용량(미터법)
DS2127-003030 mL
DS2127-0250250 mL
DS2127-20002 L
카탈로그 번호 DS2127-0030
제품 가격(KRW)
-
용량(미터법):
30 mL

Perform QA lot sample storage of high-valued pharmaceuticals using Thermo Scientific™ Nalgene™ Polycarbonate Validation Bottles. Small-volume bottles are molded of the same materials as larger carboys, but require a lower volume of expensive reagents to perform validation testing.

Autoclavable, transparent polycarbonate construction allows for convenient material compatibility validation.

Includes:
White polypropylene screw closure and TPE gasket

사양
용량(영국식 단위)1 oz.
마개 재질Polypropylene
마개 크기20-415
직경(미터법) 내부 넥14 mm
직경(미터법) 외부32 mm
용도(애플리케이션)Perform validation testing
높이(미터법) 마개 포함75 mm
케이스당 수30/Cs.
팩당 수30 Pk.
오토클레이브 가능Autoclavable
용량(미터법)30 mL
색상White (Translucent)
물질Polycarbonate
Mouth StyleWide
제품 유형Validation Bottle
멸균Non-sterile
Unit SizeCase of 30