Nalgene™ Polypropylene, Closure with Barbed Bulkhead Fittings, 1/4 inch
Thermo Scientific™
Nalgene™ Polypropylene, Closure with Barbed Bulkhead Fittings, 1/4 inch
For Use With: Nalgene culture vessels
Have Questions?
Catalog NumberQuantity
DS2167-0002Case of 4
Catalog number DS2167-0002
Price (MXN)
-
Quantity:
Case of 4
Use Thermo Scientific™ Nalgene™ Closures with Barbed Bulkhead Fittings to customize most Nalgene bottle and carboy closures for aseptic transfer of liquids.
  • Size 38mm-430 closures are predrilled allowing for easy, safe in-house assembly
  • Barbed at both ends to accept tubing inside and outside the container
  • Molded of white, autoclavable polypropylene, ready for in-house sterilization
Includes:
Instructions, drilling template and a set of two panel-mounted barbed fittings, acetal nuts and silicone gaskets.
Ordering Information:
Barbed bulkhead fittings also sold separately (6149-0001 and 6149-0002).
Specifications
Closure Size38-430
DescriptionClosure with Barbed Bulkhead Fitting
Diameter (English) Inner Tubing1/4 in.
For Use With (Equipment)Nalgene culture vessels
No. per Case4/Cs.
No. per Pack4 Pk.
QuantityCase of 4
MaterialPolypropylene
Product TypeClosure
Unit SizeCase of 4