Nalgene™ Autoclavable Septum Closure, Autoclavable, 38-430 size
Nalgene™ Autoclavable Septum Closure, Autoclavable, 38-430 size
Nalgene™ Autoclavable Septum Closure, Autoclavable, 38-430 size
Thermo Scientific™
Nalgene™ Autoclavable Septum Closure, Autoclavable, 38-430 size
Enable aseptic injection of reagent or sample withdrawal without compromising sterility or integrity of contents with this autoclavable septum closure.
Have Questions?
Catalog NumberThread Size (Metric)
DS2168-0384S2 mm
Catalog number DS2168-0384
Price (USD)
56.75
Each
Thread Size (Metric):
S2 mm
Price (USD)
56.75
Each
Enable aseptic injection of reagent or sample withdrawal without compromising sterility or integrity of contents with Thermo Scientific™ Nalgene™ Autoclavable Septum Closure, suitable for use with any bottle or container with 38-430 neck, including Nalgene culture vessels.
  • Use with 18-gauge or smaller needles
  • Polypropylene closure with thermoplastic elastomer septum
Specifications
AutoclavableAutoclavable
DescriptionAutoclavable Septum Closures
For Use With (Equipment)Nalgene bottles with 38-430 threaded mouth.
Neck Finish38-430
No. per Case12
Product TypeSeptum Closure
QuantityEach
Thread Size (Metric)S2 mm
ColorWhite
MaterialPolypropylene
MouthThreaded
SterilityNon-sterile
Unit SizeEach