Nalgene™ Magnetic PVDF Carboy Stirrer
Thermo Scientific™

Nalgene™ Magnetic PVDF Carboy Stirrer

For Use With: 10 or 20L Nalgene carboys with 83B closure
Have Questions?
Catalog NumberQuantity
DS2227-0020Each
Catalog number DS2227-0020
Price (MXN)
-
Quantity:
Each

Mix high volumes of media and buffer solution at low speed using the Thermo Scientific™ Nalgene™ Magnetic PVDF Carboy Stirrer. Adjusts easily to fit either a 10 or 20L Nalgene carboy with an 83B size closure. Supplied with a closure and two impellers. Eliminates the need to retrieve magnets from your solution.

Features:
  • For use with a magnetic stir plate (not included)
  • Includes closure and two impellers
  • Autoclavable
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
Closure Size83B
DescriptionStir bar D x L: 1/2 x 3 in.
For Use With (Equipment)10 or 20L Nalgene carboys with 83B closure
Length (English)3 in.
Length (Metric)588 mm
No. per Case1 Ea.
No. per Pack1 Ea.
QuantityEach
MaterialPTFE, PVDF, Stainless Steel
Product TypeStirrer
Unit SizeEach