Nalgene™ Fluorinated HDPE, Carboy with Spigot - FAQs
Nalgene™ Fluorinated HDPE, Carboy with Spigot - FAQs
View additional product information for Nalgene™ Fluorinated HDPE, Carboy with Spigot - FAQs (DS2327-0050, DS2327-0020)
1 product FAQs found
To what level are your FLPE fluorinated containers processed?
All of our fluorinated products are processed to Level 5.