Nalgene™ PPCO Dispensing Bottle with Closure: Autoclavable, 125mL - FAQs
Nalgene™ PPCO Dispensing Bottle with Closure: Autoclavable, 125mL - FAQs
View additional product information for Nalgene™ PPCO Dispensing Bottle with Closure: Autoclavable - FAQs (DS2420-0250, DS2420-0125)
1 product FAQs found
Are the Nalgene PPCO media bottles/tubes able to be irradiated?
PPCO bottles/tubes are not recommended for sterilization by radiation. Polycarbonate bottles/tubes would be a suitable alternative if radiation sterilization is desired.