Support 250mL bottles and ensure proper rotor fit with the Thermo ScientificTM NalgeneTM LDPE Centrifuge Bottle Adapter.
Have Questions?
產品號碼
No. per Pack
DS3125-0250
6
產品號碼 DS3125-0250
價格 (TWD)
-
No. per Pack:
6
Support 250mL bottles and ensure proper rotor fit with the Thermo Scientific™ Nalgene™ LDPE Centrifuge Bottle Adapter. For use with Thermo Scientific™ Sorvall™ GSA rotors..
For use with Nalgene 250ml centrifuge bottles catalog numbers 3120-0250, 3121-0250, 3122-0250, 3127-0250, 3140-0250 and 3141-0250
Adapts Nalgene 250ml centrifuge bottles to fit in Thermo Scientific Sorval GSA rotors
Molded of white, low-density polyethylene
規格
ColorWhite
DescriptionCentrifuge Bottle Adapter for 250mL bottles