Nalgene™ Sealing Caps for Oak Ridge Centrifuge Tubes
Nalgene™ Sealing Caps for Oak Ridge Centrifuge Tubes
Nalgene™ Sealing Caps for Oak Ridge Centrifuge Tubes
Nalgene™ Sealing Caps for Oak Ridge Centrifuge Tubes
Thermo Scientific™
Nalgene™ Sealing Caps for Oak Ridge Centrifuge Tubes
Use these sealing caps to ensure leakproof service of centrifuge bottles at their recommended maximum rated speeds.
Have Questions?
Alterar visualizaçãobuttonViewtableView
Número do catálogoClosure SizeFor Use With (Equipment)
DS3132-002424 mm50 mL Tubes
DS3132-002020 mm28 and 30 mL Tubes
DS3132-005858 mm250 mL Tubes
DS3132-006363 mm500 mL and 1 L Tubes
Número do catálogo DS3132-0024
Preço (BRL)
-
Closure Size:
24 mm
For Use With (Equipment):
50 mL Tubes
Ensure leakproof† service of Thermo Scientific™ Nalgene™ centrifuge bottles at their recommended maximum rated speeds by using Nalgene Sealing Caps for Oak Ridge Centrifuge Tubes. They are also recommended for use when spinning hazardous materials.
Compatibility:
  • 20 mm size – for use with Nalgene Oakridge tubes, 30ml volume
  • 24 mm size – for use with Nalgene Oakridge tubes, 35ml and 50ml volumes
  • 58 mm size - For use with Nalgene Centrifuge Bottles 3140-0250 and 3141-0250
  • 63 mm size - For use with Nalgene Centrifuge Bottles 3140-0500, 3141-0500, 3120-1000, 3120-1010, 3122-1000 and 3122-1010
Note:
Before autoclaving, set closure on top of the container without engaging the threads.
† The term leakproof applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for 2 minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.
Especificações
Closure Size24 mm
DescriptionSealing Caps for Oak Ridge Centrifuge Tubes
For Use With (Equipment)50 mL Tubes
No. per Case2
No. per Pack2
Product LineNalgene
AutoclavableAutoclavable
MaterialPolypropylene, Rubber
Product TypeCentrifuge Tube Cap
Unit SizeCase of 2