Thermo Fisher Scientific™
DataTaker DT80 Series Data Loggers
Catalog number: DT80-AL
Thermo Fisher Scientific™
DataTaker DT80 Series Data Loggers
Catalog number: DT80-AL
DataTaker DT80 series comprises a family of data loggers, from five channel data loggers to systems with hundreds of analog inputs and many digital inputs. It measures and logs analog and digital data from numerous electrical sensors or analog instruments, RTDs, thermocouples, temperature sensors, vibrating wire, strain gauges, current loops, voltages, resistance and frequencies. DataTaker models may include an Ethernet port, USB port, Modem for remote access and SMS alerts, WiFi, MODBUS connection, and may include a built-in web server, dEX, for setup and for viewing/downloading of data.
Catalog Number
DT80-AL
Unit Size
Each
Type
General Purpose Datalogger
Includes
Logger only (no accessories)
Price (USD)
Full specifications
TypeGeneral Purpose Datalogger
Display TypeLCD, Backlit, 2 line by 16 characters
Data Memory128 MB
CalibrationNot Calibrated
Alarm TypesE-mail, SMS, Physical/Digital Output
Battery Type6 V Lead Acid
IncludesLogger only (no accessories)
Channels5 to 15 Analog and 12 Digital (expandable using a CEM20 modules), high speed counter inputs and phase encoder inputs
InterfaceIncludes SDI-12, Ethernet, MODBUS, RS232, RS422, RS485, USB
IP RatingIP 54
ModelDT80-AL
Sensor TypesNumerous, including thermocouples, thermistors, temperature sensors, strain gauges and bridge sensors
Humidity RangeMaximum: 85%, non-condensing
Operating Temperature (Metric)-30°C to +70°C (reduced battery life and LCD operation outside range -15°C to +50°C)
Dimensions (L x W x H)180 x 137 x 65 mm
Weight (Metric)1.5 kg
Unit SizeEach
Catalog Number
Unit Size
Each
Type
General Purpose Datalogger
Includes
Logger and accessories
Price (USD)
Full specifications
TypeGeneral Purpose Datalogger
Display TypeLCD, Backlit, 2 line by 16 characters
Data Memory128 MB
CalibrationNot Calibrated
Alarm TypesE-mail, SMS, Physical/Digital Output
Battery Type6 V Lead Acid
IncludesLogger and accessories
Channels5 to 15 Analog and 12 Digital (expandable using a CEM20 modules), high speed counter inputs and phase encoder inputs
InterfaceIncludes SDI-12, Ethernet, MODBUS, RS232, RS422, RS485, USB
IP RatingIP 54
ModelDT80
Sensor TypesNumerous, including thermocouples, thermistors, temperature sensors, strain gauges and bridge sensors
Humidity RangeMaximum: 85%, non-condensing
Operating Temperature (Metric)-30°C to +70°C (reduced battery life and LCD operation outside range -15°C to +50°C)
Dimensions (L x W x H)180 x 137 x 65 mm
Weight (Metric)1.5 kg
Unit SizeEach
Catalog Number
Unit Size
Each
Type
General Purpose Datalogger
Includes
Logger with integrated modem and accessories
Price (USD)
Full specifications
TypeGeneral Purpose Datalogger
Display TypeLCD, Backlit, 2 line by 16 characters
Data Memory128 MB
CalibrationNot Calibrated
Alarm TypesE-mail, SMS, Physical/Digital Output
Battery TypeNone, external power required
IncludesLogger with integrated modem and accessories
Channels5 to 15 Analog and 12 Digital (expandable using a CEM20 modules), high speed counter inputs and phase encoder inputs
InterfaceIncludes SDI-12, Ethernet, MODBUS, RS232, RS422, RS485, USB
IP RatingIP 54
ModelDT80M
Sensor TypesNumerous, including thermocouples, thermistors, temperature sensors, strain gauges and bridge sensors
Humidity RangeMaximum: 85%, non-condensing
Operating Temperature (Metric)-30°C to +70°C (reduced battery life and LCD operation outside range -15°C to +50°C)
Dimensions (L x W x H)180 x 137 x 65 mm
Weight (Metric)1.5 kg
Unit SizeEach
Catalog Number
Unit Size
Each
Type
General Purpose Datalogger
Includes
Logger with integrated Wifi and accessories
Price (USD)
Full specifications
TypeGeneral Purpose Datalogger
Display TypeLCD, Backlit, 2 line by 16 characters
Data Memory128 MB
CalibrationNot Calibrated
Alarm TypesE-mail, SMS, Physical/Digital Output
Battery Type6 V Lead Acid
IncludesLogger with integrated Wifi and accessories
Channels5 to 15 Analog and 12 Digital (expandable using a CEM20 modules), high speed counter inputs and phase encoder inputs
InterfaceIncludes SDI-12, Ethernet, MODBUS, RS232, RS422, RS485, USB
IP RatingIP 54
ModelDT80W
Sensor TypesNumerous, including thermocouples, thermistors, temperature sensors, strain gauges and bridge sensors
Humidity RangeMaximum: 85%, non-condensing
Operating Temperature (Metric)-30°C to +70°C (reduced battery life and LCD operation outside range -15°C to +50°C)
Dimensions (L x W x H)180 x 137 x 65 mm
Weight (Metric)1.5 kg
Unit SizeEach
Catalog Number
Unit Size
Each
Type
Geotechnical logger supporting vibrating wire strain gauges
Includes
Logger only (no accessories)
Price (USD)
Full specifications
TypeGeotechnical logger supporting vibrating wire strain gauges
Display TypeLCD, Backlit, 2 line by 16 characters
Data Memory128 MB
CalibrationNot Calibrated
Alarm TypesE-mail, SMS, Physical/Digital Output
Battery TypeNone, external power required
IncludesLogger only (no accessories)
Channels5 to 15 Analog and 12 Digital (expandable using a CEM20 modules), high speed counter inputs and phase encoder inputs
InterfaceIncludes SDI-12, Ethernet, MODBUS, RS232, RS422, RS485, USB
IP RatingIP 54
ModelDT80G-AL
Sensor TypesNumerous, including vibrating wire sensors, thermocouples, thermistors, temperature sensors, strain gauges and bridge sensors
Humidity RangeMaximum: 85%, non-condensing
Operating Temperature (Metric)-30°C to +70°C (reduced battery life and LCD operation outside range -15°C to +50°C)
Dimensions (L x W x H)180 x 137 x 65 mm
Weight (Metric)1.5 kg
Unit SizeEach
Catalog Number
Unit Size
Each
Type
Geotechnical logger supporting vibrating wire strain gauges
Includes
Logger and accessories
Price (USD)
Full specifications
TypeGeotechnical logger supporting vibrating wire strain gauges
Display TypeLCD, Backlit, 2 line by 16 characters
Data Memory128 MB
CalibrationNot Calibrated
Alarm TypesE-mail, SMS, Physical/Digital Output
Battery TypeNone, external power required
IncludesLogger and accessories
Channels5 to 15 Analog and 12 Digital (expandable using a CEM20 modules), high speed counter inputs and phase encoder inputs
InterfaceIncludes SDI-12, Ethernet, MODBUS, RS232, RS422, RS485, USB
IP RatingIP 54
ModelDT80G
Sensor TypesNumerous, including vibrating wire sensors, thermocouples, thermistors, temperature sensors, strain gauges and bridge sensors
Humidity RangeMaximum: 85%, non-condensing
Operating Temperature (Metric)-30°C to +70°C (reduced battery life and LCD operation outside range -15°C to +50°C)
Dimensions (L x W x H)180 x 137 x 65 mm
Weight (Metric)1.5 kg
Unit SizeEach
Showing 6 of 6
Catalog NumberSpecificationsUnit SizeTypeIncludesPrice (USD)
DT80-ALFull specifications
EachGeneral Purpose DataloggerLogger only (no accessories)Contact Us ›
TypeGeneral Purpose Datalogger
Display TypeLCD, Backlit, 2 line by 16 characters
Data Memory128 MB
CalibrationNot Calibrated
Alarm TypesE-mail, SMS, Physical/Digital Output
Battery Type6 V Lead Acid
IncludesLogger only (no accessories)
Channels5 to 15 Analog and 12 Digital (expandable using a CEM20 modules), high speed counter inputs and phase encoder inputs
InterfaceIncludes SDI-12, Ethernet, MODBUS, RS232, RS422, RS485, USB
IP RatingIP 54
ModelDT80-AL
Sensor TypesNumerous, including thermocouples, thermistors, temperature sensors, strain gauges and bridge sensors
Humidity RangeMaximum: 85%, non-condensing
Operating Temperature (Metric)-30°C to +70°C (reduced battery life and LCD operation outside range -15°C to +50°C)
Dimensions (L x W x H)180 x 137 x 65 mm
Weight (Metric)1.5 kg
Unit SizeEach
DT80Full specifications
EachGeneral Purpose DataloggerLogger and accessoriesContact Us ›
TypeGeneral Purpose Datalogger
Display TypeLCD, Backlit, 2 line by 16 characters
Data Memory128 MB
CalibrationNot Calibrated
Alarm TypesE-mail, SMS, Physical/Digital Output
Battery Type6 V Lead Acid
IncludesLogger and accessories
Channels5 to 15 Analog and 12 Digital (expandable using a CEM20 modules), high speed counter inputs and phase encoder inputs
InterfaceIncludes SDI-12, Ethernet, MODBUS, RS232, RS422, RS485, USB
IP RatingIP 54
ModelDT80
Sensor TypesNumerous, including thermocouples, thermistors, temperature sensors, strain gauges and bridge sensors
Humidity RangeMaximum: 85%, non-condensing
Operating Temperature (Metric)-30°C to +70°C (reduced battery life and LCD operation outside range -15°C to +50°C)
Dimensions (L x W x H)180 x 137 x 65 mm
Weight (Metric)1.5 kg
Unit SizeEach
DT80MFull specifications
EachGeneral Purpose DataloggerLogger with integrated modem and accessoriesContact Us ›
TypeGeneral Purpose Datalogger
Display TypeLCD, Backlit, 2 line by 16 characters
Data Memory128 MB
CalibrationNot Calibrated
Alarm TypesE-mail, SMS, Physical/Digital Output
Battery TypeNone, external power required
IncludesLogger with integrated modem and accessories
Channels5 to 15 Analog and 12 Digital (expandable using a CEM20 modules), high speed counter inputs and phase encoder inputs
InterfaceIncludes SDI-12, Ethernet, MODBUS, RS232, RS422, RS485, USB
IP RatingIP 54
ModelDT80M
Sensor TypesNumerous, including thermocouples, thermistors, temperature sensors, strain gauges and bridge sensors
Humidity RangeMaximum: 85%, non-condensing
Operating Temperature (Metric)-30°C to +70°C (reduced battery life and LCD operation outside range -15°C to +50°C)
Dimensions (L x W x H)180 x 137 x 65 mm
Weight (Metric)1.5 kg
Unit SizeEach
DT80WFull specifications
EachGeneral Purpose DataloggerLogger with integrated Wifi and accessoriesContact Us ›
TypeGeneral Purpose Datalogger
Display TypeLCD, Backlit, 2 line by 16 characters
Data Memory128 MB
CalibrationNot Calibrated
Alarm TypesE-mail, SMS, Physical/Digital Output
Battery Type6 V Lead Acid
IncludesLogger with integrated Wifi and accessories
Channels5 to 15 Analog and 12 Digital (expandable using a CEM20 modules), high speed counter inputs and phase encoder inputs
InterfaceIncludes SDI-12, Ethernet, MODBUS, RS232, RS422, RS485, USB
IP RatingIP 54
ModelDT80W
Sensor TypesNumerous, including thermocouples, thermistors, temperature sensors, strain gauges and bridge sensors
Humidity RangeMaximum: 85%, non-condensing
Operating Temperature (Metric)-30°C to +70°C (reduced battery life and LCD operation outside range -15°C to +50°C)
Dimensions (L x W x H)180 x 137 x 65 mm
Weight (Metric)1.5 kg
Unit SizeEach
DT80G-ALFull specifications
EachGeotechnical logger supporting vibrating wire strain gaugesLogger only (no accessories)Contact Us ›
TypeGeotechnical logger supporting vibrating wire strain gauges
Display TypeLCD, Backlit, 2 line by 16 characters
Data Memory128 MB
CalibrationNot Calibrated
Alarm TypesE-mail, SMS, Physical/Digital Output
Battery TypeNone, external power required
IncludesLogger only (no accessories)
Channels5 to 15 Analog and 12 Digital (expandable using a CEM20 modules), high speed counter inputs and phase encoder inputs
InterfaceIncludes SDI-12, Ethernet, MODBUS, RS232, RS422, RS485, USB
IP RatingIP 54
ModelDT80G-AL
Sensor TypesNumerous, including vibrating wire sensors, thermocouples, thermistors, temperature sensors, strain gauges and bridge sensors
Humidity RangeMaximum: 85%, non-condensing
Operating Temperature (Metric)-30°C to +70°C (reduced battery life and LCD operation outside range -15°C to +50°C)
Dimensions (L x W x H)180 x 137 x 65 mm
Weight (Metric)1.5 kg
Unit SizeEach
DT80GFull specifications
EachGeotechnical logger supporting vibrating wire strain gaugesLogger and accessoriesContact Us ›
TypeGeotechnical logger supporting vibrating wire strain gauges
Display TypeLCD, Backlit, 2 line by 16 characters
Data Memory128 MB
CalibrationNot Calibrated
Alarm TypesE-mail, SMS, Physical/Digital Output
Battery TypeNone, external power required
IncludesLogger and accessories
Channels5 to 15 Analog and 12 Digital (expandable using a CEM20 modules), high speed counter inputs and phase encoder inputs
InterfaceIncludes SDI-12, Ethernet, MODBUS, RS232, RS422, RS485, USB
IP RatingIP 54
ModelDT80G
Sensor TypesNumerous, including vibrating wire sensors, thermocouples, thermistors, temperature sensors, strain gauges and bridge sensors
Humidity RangeMaximum: 85%, non-condensing
Operating Temperature (Metric)-30°C to +70°C (reduced battery life and LCD operation outside range -15°C to +50°C)
Dimensions (L x W x H)180 x 137 x 65 mm
Weight (Metric)1.5 kg
Unit SizeEach
Showing 6 of 6
The DataTaker DT80 series comprises a family of industrial data loggers, ranging from five channel data loggers to systems with hundreds of analog inputs and many digital inputs.
The DataTaker DT80 series measures and logs analog & digital data from numerous electrical sensors or analog instruments, RTDs, thermocouples, temperature sensors, strain gauges, current loops, voltages, resistance and frequencies.
DataTaker DT80 models include an Ethernet port, USB port and a built-in web server, dEX for setup and for viewing/downloading of data. In addition:
  • DT80M includes an integrated modem
  • DT80W and DT80GW include integrated WiFi
DataTaker DT80G is specifically suited to geotechnical applications such as slope stability, subsidence, dam wall monitoring, tunnel and mining excavation, ground water, tunnel wall monitoring and site assessment. DT80G supports vibrating wire sensors and other geotechnical sensors.
Other Features:
  • Analog input channels set up as binary logic level inputs.
  • Digital inputs recording ON/OFF states of contacts, or counting pulses over a desired period.
  • Alarm channels and digital outputs to raise alarms (email / SMS / physical digital outputs) or control external processes.
  • Readings may be returned directly or via internal calculations based on a single channel or multiple channels.
  • Internal memory holds ∼10 million readings.
  • Connection to the host computer via Ethernet, USB, and serial Host Port.
  • Separate USB port for USB flash drives to be used for extra storage, or automatic unloading of data from the logger.
  • Built-in battery or auxiliary battery option.
  • Most data logging configurations are easily set up using the graphical web-based dEX interface and a standard web browser.
  • For more complex configurations, a script can be developed using the logger's own scripting language.