DHX™ Heat Exchanger BioProcess Container (BPC), ASI™ 26/77 film, ReadyMate connections, two 6-L BPCs
Thermo Scientific™

DHX™ Heat Exchanger BioProcess Container (BPC), ASI™ 26/77 film, ReadyMate connections, two 6-L BPCs

The Thermo Scientific™ DHX™ Heat Exchanger Single-Use BioProcess Container (BPC) is a 2D system consisting of two 6 L BPCs,深入閱讀
Have Questions?
產品號碼Quantity
DX00007I
亦稱為 DX00007-I
2 x 6 L
產品號碼 DX00007I
亦稱為 DX00007-I
價格 (HKD)
-
Quantity:
2 x 6 L
The Thermo Scientific™ DHX™ Heat Exchanger Single-Use BioProcess Container (BPC) is a 2D system consisting of two 6 L BPCs, without drain. It is composed of Thermo Scientific™ ASI™ 26/77 Polyethylene Film, with interconnecting TPE tubing and ReadyMate DAC end connections. The DHX Heat Exchanger and DHX BPC each features a serpentine pattern, creating a sterile fluid path for the process fluid. The BPC fits tightly between the stainless steel DHX Heat Exchanger plate system, and after loading, no further operator interaction is required. The customer has the option of utilizing up to 4 BPCs in the DHX unit.

Features
• Eliminates post-use cleaning steps required with reusable containers, helping to reduce cross-contamination risks
• Provides sterile processing environment without the cost and time of clean-in-place or steam-in-place sterilization (CIP/SIP)
• Minimizes bioburden by using the DHX Heat Exchanger unit and DHX BPCs to address weak points in a CIP system or sanitation process (typically required for stainless steel equipment)
• Reduces maintenance and life cycle cost due to its unique design
• Increases volume capacity via parallel connection between DHX units

Applications
• cGMP commercial and clinical biotherapeutics, vaccines, and other biologic processes
• Upstream applications, including media hold, harvest cooling, and harvest hold
• Downstream applications, including harvest hold, buffers, protein purification, and bulk drug substance

規格
DimensionsEach BPC: 62.2 cm x 61.9 cm (24.5 in. x 24.4 in.)
Film DescriptionASI™ 26/77
For Use With (Equipment)DHX1001
MaterialPolyethylene
Outer PackagingTwo 4-mil. poly bags
Port DetailReadyMate DAC connectors
Product LineDHX
Protective Inner PackagingProtective material around rough areas
Quantity2 x 6 L
Shipping ConditionRoom Temperature
Sterilization MethodGamma Irradiated
Capacity (Metric)12 L
Tubing StyleTPE
Unit SizeEach
內容物與存放
Store at room temperature.