DHX™ Heat Exchanger BioProcess Container (BPC), ASI™ 26/77 film, ReadyMate connections, drain ports, two 6-L BPCs
Thermo Scientific™

DHX™ Heat Exchanger BioProcess Container (BPC), ASI™ 26/77 film, ReadyMate connections, drain ports, two 6-L BPCs

The Thermo Scientific™ DHX™ Heat Exchanger Single-Use BioProcess Container (BPC) is a 2D system consisting of two 6 L BPCs,Read more
Have Questions?
Catalog NumberQuantity
DX00011I
also known as DX00011-I
2 x 6 L
Catalog number DX00011I
also known as DX00011-I
Price (MXN)
-
Quantity:
2 x 6 L
The Thermo Scientific™ DHX™ Heat Exchanger Single-Use BioProcess Container (BPC) is a 2D system consisting of two 6 L BPCs, with drain. It is composed of Thermo Scientific™ ASI™ 26/77 Polyethylene Film, with interconnecting TPE tubing and ReadyMate DAC end connections. The DHX Heat Exchanger and DHX BPC each features a serpentine pattern, creating a sterile fluid path for the process fluid. The BPC fits tightly between the stainless steel DHX Heat Exchanger plate system, and after loading, no further operator interaction is required. The customer has the option of utilizing up to 4 BPCs in the DHX unit.

Features
• Eliminates post-use cleaning steps required with reusable containers, helping to reduce cross-contamination risks
• Provides sterile processing environment without the cost and time of clean-in-place or steam-in-place sterilization (CIP/SIP)
• Minimizes bioburden by using the DHX Heat Exchanger unit and DHX BPCs to address weak points in a CIP system or sanitation process (typically required for stainless steel equipment)
• Reduces maintenance and life cycle cost due to its unique design
• Increases volume capacity via parallel connection between DHX units

Applications
• cGMP commercial and clinical biotherapeutics, vaccines, and other biologic processes
• Upstream applications, including media hold, harvest cooling, and harvest hold
• Downstream applications, including harvest hold, buffers, protein purification, and bulk drug substance

Specifications
DimensionsEach BPC: 62.2 cm x 61.9 cm (24.5 in. x 24.4 in.)
Film DescriptionASI™ 26/77
For Use With (Equipment)DHX1001
MaterialPolyethylene
Outer PackagingTwo 4-mil. poly bags
Port DetailReadyMate DAC connectors and drain port
Product LineDHX
Protective Inner PackagingProtective material around rough areas
Quantity2 x 6 L
Shipping ConditionRoom Temperature
Sterilization MethodGamma Irradiated
Capacity (Metric)12 L
Tubing StyleTPE, ID x OD: 0.64 x 12.7 cm (0.25 in. x 0.5 in.)
Unit SizeEach
Contents & Storage
Store at room temperature.