Ethanediol, Extra Pure, SLR, Fisher Chemical™
Ethanediol, Extra Pure, SLR, Fisher Chemical™
Ethanediol, Extra Pure, SLR, Fisher Chemical™
Fisher Chemical™

Ethanediol, Extra Pure, SLR, Fisher Chemical™

C2H6O2, CAS Number-107-21-1, ethylene alcohol, glycol alcohol, 2-hydroxyethanol, 1,2-ethanediol, glycol, fridex, monoethylene glycol, tescol, 1,2-dihydroxyethane, ethylene glycol, 2.5L, 196 deg.C, CHEBI:30742, Colorless, 62.06g/mol, LYCAIKOWRPUZTN-UHFFFAOYSA-N, ethane-1,2-diol, -13 deg.C, 2885 | CAS: 107-21-1 | C2H6O2 | 62.068 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
2.5 L
Packaging:
Amber glass bottle
Catalog number E-0350-15
also known as E/0350/15
Price (USD)
-
Quantity:
1 L
Packaging:
Amber glass bottle
Request bulk or custom format
Specifications
CAS107-21-1
ChEBICHEBI:30742
ColorColorless
Formula Weight62.06g/mol
IUPAC Nameethane-1,2-diol
View more
C2H6O2, CAS Number-107-21-1, ethylene alcohol, glycol alcohol, 2-hydroxyethanol, 1,2-ethanediol, glycol, fridex, monoethylene glycol, tescol, 1,2-dihydroxyethane, ethylene glycol, 2.5L, 196 deg.C, CHEBI:30742, Colorless, 62.06g/mol, LYCAIKOWRPUZTN-UHFFFAOYSA-N, ethane-1,2-diol, -13 deg.C, 2885