Ethanol Absolute 99.8+%, Certified AR for Analysis, meets analytical specification of Ph.Eur., BP, Fisher Chemical™
Ethanol Absolute 99.8+%, Certified AR for Analysis, meets analytical specification of Ph.Eur., BP, Fisher Chemical™
Ethanol Absolute 99.8+%, Certified AR for Analysis, meets analytical specification of Ph.Eur., BP, Fisher Chemical™
Ethanol Absolute 99.8+%, Certified AR for Analysis, meets analytical specification of Ph.Eur., BP, Fisher Chemical™
Fisher Chemical™

Ethanol Absolute 99.8+%, Certified AR for Analysis, meets analytical specification of Ph.Eur., BP, Fisher Chemical™

C2H6O, CAS Number-64-17-5, 2.5L, 78 deg.C, CHEBI:16236, Colorless, 46.07g/mol, LFQSCWFLJHTTHZ-UHFFFAOYSA-N, ethanol, -114 deg.C, 3568, 46.069, Amber glass bottle, Liquid, 702, CCO | CAS: 64-17-5 | C2H6O | 46.069 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
2.5 L
5 L
Packaging:
View all
Amber glass bottle
Catalog number E-0650DF-17
also known as E/0650DF/17
Price (USD)
-
Quantity:
2.5 L
Packaging:
Amber glass bottle
Request bulk or custom format
Specifications
Boiling Point78°C
CAS64-17-5
ChEBICHEBI:16236
ColorColorless
Formula Weight46.07g/mol
View more
C2H6O, CAS Number-64-17-5, 2.5L, 78 deg.C, CHEBI:16236, Colorless, 46.07g/mol, LFQSCWFLJHTTHZ-UHFFFAOYSA-N, ethanol, -114 deg.C, 3568, 46.069, Amber glass bottle, Liquid, 702, CCO