Ethyl Methyl Ketone, Extra Pure, SLR, Fisher Chemical™
Ethyl Methyl Ketone, Extra Pure, SLR, Fisher Chemical™
Ethyl Methyl Ketone, Extra Pure, SLR, Fisher Chemical™
Fisher Chemical™

Ethyl Methyl Ketone, Extra Pure, SLR, Fisher Chemical™

C4H8O, CAS Number-78-93-3, methyl ethyl ketone, methylethylketone, methyl acetone, 2-butanone, methylethyl ketone, meetco, butanone, 3-butanone, ethyl methyl ketone, ethylmethylketon, 2.5L, 80 deg.C, CHEBI:28398, Colorless, 72.11g/mol, ZWEHNKRNPOVVGH-UHFFFAOYSA-N, butan-2-one, -87 deg.C, 11648
Have Questions?
變更視圖buttonViewtableView
Quantity:
500 mL
1 L
2.5 L
Packaging:
Amber glass bottle
產品號碼 E-1450-15
亦稱為 E/1450/15
價格 (TWD)
-
Quantity:
1 L
Packaging:
Amber glass bottle
Request bulk or custom format
規格
CAS78-93-3
ChEBICHEBI:28398
ColorColorless
Formula Weight72.11g/mol
IUPAC Namebutan-2-one
檢視更多
C4H8O, CAS Number-78-93-3, methyl ethyl ketone, methylethylketone, methyl acetone, 2-butanone, methylethyl ketone, meetco, butanone, 3-butanone, ethyl methyl ketone, ethylmethylketon, 2.5L, 80 deg.C, CHEBI:28398, Colorless, 72.11g/mol, ZWEHNKRNPOVVGH-UHFFFAOYSA-N, butan-2-one, -87 deg.C, 11648