Ethyl Methyl Ketone, Certified AR for Analysis, Fisher Chemical™
Ethyl Methyl Ketone, Certified AR for Analysis, Fisher Chemical™
Ethyl Methyl Ketone, Certified AR for Analysis, Fisher Chemical™
Fisher Chemical™

Ethyl Methyl Ketone, Certified AR for Analysis, Fisher Chemical™

Have Questions?
Catalog NumberQuantityPackaging
E-1455-17
also known as E/1455/17
2.5 LAmber glass bottle
Catalog number E-1455-17
also known as E/1455/17
Price (TWD)
-
Quantity:
2.5 L
Packaging:
Amber glass bottle
Request bulk or custom format
Specifications
CAS78-93-3
ChEBICHEBI:28398
ColorColorless
Formula Weight72.11g/mol
IUPAC Namebutan-2-one
View more
Ethyl Methyl Ketone, Certified AR for Analysis