AlbuMax II 20% Solution BSA, IRR, 1 L
Gibco™

AlbuMax II 20% Solution BSA, IRR, 1 L

Lipid-rich bovine serum albumin for cell culture. It is chromatographically purified and has an IgG content ≤1.0%. 20% solution with additional irradiation.
Have Questions?
보기 방식 변경buttonViewtableView
카탈로그 번호수량
E003001PK1 L
E003001PR5 L
카탈로그 번호 E003001PK
제품 가격(KRW)
-
수량:
1 L
Lipid-rich bovine serum albumin for cell culture. It is chromatographically purified and has an IgG content ≤1.0%. 20% solution with additional irradiation.
For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.
사양
세포 유형Mammalian Cells
제품 유형BSA
수량1 L
형태Liquid
Unit SizeEach