BSA Fraction V UL IGG
BSA Fraction V UL IGG
Gibco™
BSA Fraction V UL IGG
Ultra-low IgG bovine serum albumin for cell culture. It is chromatographically purified and has an IgG content ≤4ppm. Lyophilized powder.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
E014000IL25 g
E014000IM100 g
E014000IP1 kg
E014000IQ5 kg
Catalog number E014000IL
Price (MXN)
-
Request A Quote
Quantity:
25 g
Ultra-low-IgG bovine serum albumin for cell culture. It is chromatographically purified and has an IgG content ≤4 ppm. Lyophilized powder.
For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.
Specifications
Cell TypeMammalian Cells
Product TypeBSA
Quantity25 g
FormSolid
Unit SizeEach