EnzChek® Direct Phospholipase C Assay kit for 10 plates - FAQs
EnzChek® Direct Phospholipase C Assay kit for 10 plates - FAQs
View additional product information for EnzChek® Direct Phospholipase C Assay kit for 10 plates - FAQs (E10216)
1 product FAQs found
What is the excitation/emission maxima of the reaction product when using the EnzChek Direct Phospholipase C Assay kit?
The fluorescence excitation/emission maxima is 509/516 nm.