EnzChek™ Phospholipase A1 Assay Kit
Invitrogen™

EnzChek™ Phospholipase A1 Assay Kit

このキットは、ホスホリパーゼA1(PLA1)(A10070)に対して当社のスタンドアロンのアッセイを実施し、必要な試薬を組み合わせて、精製酵素調製物および細胞ライセートの活性をモニタリングする完全な96ウェルマイクロプレートアッセイを2~10回実行します。これらのプロセスに関連する細胞シグナル伝達、脂質代謝詳細を見る
製品番号(カタログ番号)数量
E10219200回の反応、反応200回
製品番号(カタログ番号) E10219
価格(JPY)
37,100
キャンペーン価格
Ends: 25-Sep-2026
61,900
割引額 24,800 (40%)
Each
数量:
200回の反応、反応200回
このキットは、ホスホリパーゼA1(PLA1)(A10070)に対して当社のスタンドアロンのアッセイを実施し、必要な試薬を組み合わせて、精製酵素調製物および細胞ライセートの活性をモニタリングする完全な96ウェルマイクロプレートアッセイを2~10回実行します。これらのプロセスに関連する細胞シグナル伝達、脂質代謝、炎症反応、および病理学的疾患におけるホスホリパーゼの重要性から、蛍光ベースの活性モニタリング手法への需要が高まっています。特に、血漿および内皮に存在するホスホリパーゼは、循環LDLおよびHDLの粒子を摂動させ、アテローム生成に促進的な形態を形成します。最近のエビデンスでは、これらのリパーゼと、アルツハイマー病などのいくつかの重度神経変性疾患の進行との関連が明らかになっています。Molecular Probes’蛍光ホスホリパーゼA1基質は、精製酵素調製物、細胞ライセート、および生細胞におけるホスホリパーゼA1(PLA1)の連続モニタリングを実施できるように設計されています。PLA-1は、既存の2種類の試薬を2つの方法で改善します。1つはPLA-1に対する特異性があることで、もう1つは初期のバックグラウンドノイズを低減することでアッセイの品質が向上していることです。
研究用にのみ使用できます。診断用には使用いただけません。
仕様
検出法蛍光強度
数量200回の反応、反応200回
出荷条件室温
基質特性脂質ベースの基質、化学基質
基質タイプホスホリパーゼ基質
標的酵素ホスホリパーゼ
使用対象(アプリケーション)ホスホリパーゼA1アッセイ
製品ラインEnzChek、Molecular Probes
製品タイプホスホリパーゼアッセイ
Unit SizeEach
組成および保存条件

1キットキットは-20°C未満で保管し、湿気を避け、光から保護してください

以下のプロトコルで示されているように、ウェルあたり100 μLの96ウェルマイクロプレートで200回の反応に十分な材料、またはウェルあたり25 μL以下の384低容量マイクロプレートで400回の反応に十分な材料が入っています。

よくあるご質問(FAQ)

What is the LoD for the EnzChek Phospholipase A1 Assay Kit (Cat. No. E10219)?

The EnzChek Phospholipase A1 Assay Kit (Cat. No. E10219) can detect PLA1 at 0.04 U/mL or lower.

引用および参考文献 (3)

引用および参考文献
Abstract
Crystal structure of the predicted phospholipase LYPLAL1 reveals unexpected functional plasticity despite close relationship to acyl protein thioesterases.
Authors:Bürger M, Zimmermann TJ, Kondoh Y, Stege P, Watanabe N, Osada H, Waldmann H, Vetter IR,
Journal:J Lipid Res
PubMed ID:22052940
'Sequence homology indicates the existence of three human cytosolic acyl protein thioesterases, including APT1 that is known to depalmitoylate H- and N-Ras. One of them is the lysophospholipase-like 1 (LYPLAL1) protein that on the one hand is predicted to be closely related to APT1 but on the other hand might ... More
Furin Is the Primary in Vivo Convertase of Angiopoietin-like 3 and Endothelial Lipase in Hepatocytes.
Authors:Essalmani R, Susan-Resiga D, Chamberland A, Asselin MC, Canuel M, Constam D, Creemers JW, Day R, Gauthier D, Prat A, Seidah NG,
Journal:
PubMed ID:23918928
'The proprotein convertases (PCs) furin, PC5/6, and PACE4 exhibit unique and/or complementary functions. Their knock-out (KO) in mice resulted in strong and specific phenotypes demonstrating that, in vivo, these PCs are unique and essential during development. However, they also exhibit redundant functions. Liver angiopoietin-like 3 (ANGPTL3) inhibits lipolysis by binding ... More
Measurement of the phospholipase activity of endothelial lipase in mouse plasma.
Authors:Basu D, Lei X, Josekutty J, Hussain MM, Jin W,
Journal:J Lipid Res
PubMed ID:23103358
Endothelial lipase (EL) is a major negative regulator of plasma HDL levels in mice, rabbits, and most probably, humans. Although this regulatory function is critically dependent on EL's hydrolysis of HDL phospholipids, as yet there is no phospholipase assay specific for EL in plasma. We developed such an assay for ... More