Thermo Scientific™
RA Lamb SlimLine Digital Hotplate, 85H x 176W x 515mm D
 Related applications:
Anatomical Pathology
Drive off water and organic or flammable solvents used in microscopically mounting media with the Thermo Scientific™ RA Lamb SlimLine Digital Hotplate. With a usable heated area of 480 x 175mm providing a maximum capacity of 38 standard 76 x 26mm slides, the hotplate operates over a 40-95°C temperature range.
Error loading your content!
  Catalog number Price ({{currency}}) Availability Qty
{{product.sku}}
also known as {{product.formattedSku}} 
{{product.availability.message}} Add To Cart
{{addItemToList.successMessage}} {{addItemToList.listName}}
Back to top
Description
  • Holds two rows of 18 slides.
  • Epoxy coated steel lower casing.
  • Aluminum top with matte-black epoxy coating.
  • Includes four removable slide rods.
  • Additional use includes warming wax embedded specimens retained in conductive, leak proof trays.
Documents
Manuals & protocols