EnzChek™ Reverse Transcriptase Assay Kit
Invitrogen™

EnzChek™ Reverse Transcriptase Assay Kit

The EnzChek Reverse Transcriptase Assay Kit is a convenient, efficient, and inexpensive assay for measuring reverse transcriptase activity and screening reverse transcriptase inhibitors.

Have Questions?
Catalog NumberQuantity
E220641000 Assays
Catalog number E22064
Price (USD)
724.00
Each
Quantity:
1000 Assays
Request bulk or custom format
Price (USD)
724.00
Each

The EnzChek Reverse Transcriptase Assay Kit is a convenient, efficient, and inexpensive assay for measuring reverse transcriptase activity and screening reverse transcriptase inhibitors. In less than an hour, samples can be read in a fluorometer or microplate reader with filter sets appropriate for fluorescein (FITC), GFP, or Alexa Fluor 488.

The EnzChek Reverse Transcriptase Assay Kit is a convenient, efficient, and inexpensive assay for measuring reverse transcriptase activity and screening reverse transcriptase inhibitors. In less than an hour, samples can be read in a fluorometer or microplate reader with filter sets appropriate for fluorescein (FITC), GFP, or Alexa Fluor 488.

Features of the EnzChek Reverse Transcriptase Assay Kit include:

  • Detect as little as 0.02 units of HIV-1 reverse transcriptase
  • Large dynamic assay range, detect up to a 50-fold linear range
  • Simple to use assay, amenable for automated high-throughput screening applications

The EnzChek Reverse Transcriptase Assay Kit uses the PicoGreen reagent, which preferentially detects dsDNA or RNA-DNA heteroduplexes over single-stranded nucleic acids or free nucleotides. In this assay, the reverse transcriptase activity in a biological sample generates long RNA-DNA heteroduplexes from a mixture of a long poly(A) template, an oligo-dT primer, and dTTP. The RNA-DNA heteroduplexes formed are then detected by the PicoGreen reagent.

Reverse transcriptase is an enzyme that functions as an RNA-dependent DNA-polymerase to synthesize DNA from an RNA template. Retroviruses such as HIV use reverse transcriptase activity to make a DNA copy of their viral RNA to insert into a host cell genome. Reverse transcriptase enzymes are also used in various cell functions, such as the enzyme telomerase, which maintains telomere length and chromosome stability and has a role in cancer and aging. In molecular biology, reverse transcriptase function is utilized to generate complementary DNA (cDNA) libraries from mRNA and quantify mRNA expression through amplification in RT-PCR.

The EnzChek Reverse Transcriptase Assay Kit provides a simple method to determine reverse transcriptase activity that is useful in the study of retroviruses and screening reverse transcriptase inhibitors, which can function as therapeutic drugs to target retroviral diseases. Because this method is faster and less expensive than standard isotopic assays or immunoassays, it is suitable for testing large numbers of biological samples, enabling automated high-throughput screening of reverse transcriptase inhibitors.

For Research Use Only. Not for use in diagnostic procedures.
Specifications
Detection MethodFluorescence
Emission Wavelength523 nm
Excitation Wavelength502 nm
Quantity1000 Assays
Shipping ConditionRoom Temperature
Sufficient For1000 Assays
Target EnzymeReverse Transcriptase
Emission Wavelength Range510 to 525 nm
Excitation Wavelength Range495 to 510 nm
For Use With (Application)Enzyme and Metabolic Activity
For Use With (Equipment)Microplate Reader
Product LineEnzChek
Product TypeEnzChek Assay Kit
Unit SizeEach
Contents & Storage
Upon receipt, the kit should be stored at –20°C and protected from light. The 20X TE buffer and the lambda DNA standard are best stored at 4°C; however, either may be frozen for long term storage. Stored properly, the kit's components should remain stable for at least six months. Allow reagents to warm to room temperature before opening the vials.

Frequently asked questions (FAQs)

With the EnzChek Reverse Transcriptase Assay Kit, how do I expand the dynamic range?

To expand the upper end of the PicoGreen’s dynamic range, simply increase dye concentration. By doubling the concentration, the dynamic range will double, etc. Keep in mind, however that increasing the dye concentration sacrifices the lower end of the assay due to increasing background fluorescence.

Does the assay EnzChek Reverse Transcriptase Assay Kit (Cat. No. E22064) work on retroviruses other than HIV?

The assay should work with any reverse transcriptase.
Note: This is a solution reaction, so the reverse transcriptase needs to be available in solution to work, not encapsulated.

Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.

Citations & References (4)

Citations & References
Abstract
Solution structural dynamics of HIV-1 reverse transcriptase heterodimer.
Authors:Seckler JM, Howard KJ, Barkley MD, Wintrode PL,
Journal:Biochemistry
PubMed ID:19594135
'Crystal structures and simulations suggest that conformational changes are critical for the function of HIV-1 reverse transcriptase. The enzyme is an asymmetric heterodimer of two subunits, p66 and p51. The two subunits have the same N-terminal sequence, with the p51 subunit lacking the C-terminal RNase H domain. We used hydrogen ... More
Apobec3 encodes Rfv3, a gene influencing neutralizing antibody control of retrovirus infection.
Authors:Santiago ML, Montano M, Benitez R, Messer RJ, Yonemoto W, Chesebro B, Hasenkrug KJ, Greene WC,
Journal:Science
PubMed ID:18772436
Recovery from Friend virus 3 (Rfv3) is a single autosomal gene encoding a resistance trait that influences retroviral neutralizing antibody responses and viremia. Despite extensive research for 30 years, the molecular identity of Rfv3 has remained elusive. Here, we demonstrate that Rfv3 is encoded by Apobec3. Apobec3 maps to the ... More
Importance of SARS-CoV spike protein Trp-rich region in viral infectivity.
Authors:Lu Y, Neo TL, Liu DX, Tam JP,
Journal:Biochem Biophys Res Commun
PubMed ID:18424264
SARS-CoV entry is mediated by spike glycoprotein. During the viral and host cellular membrane fusion, HR1 and HR2 form 6-helix bundle, positioning the fusion peptide closely to the C-terminal region of ectodomain to drive apposition and subsequent membrane fusion. Connecting to the HR2 region is a Trp-rich region which is ... More
Evolution of subtype C HIV-1 Env in a slowly progressing Zambian infant.
Authors:Zhang H, Hoffmann F, He J, He X, Kankasa C, Ruprecht R, West JT, Orti G, Wood C,
Journal:Retrovirology
PubMed ID:16274482
BACKGROUND: Given the high prevalence of mother to child infection, the development of a better understanding of African subtype C HIV-1 transmission and natural evolution is of significant importance. In this study, we genotypically and phenotypically characterized subtype C viruses isolated over a 67-month follow-up period from an in utero-infected ... More