What is a "supershift"?
A supershift assay is a method for positively identifying a protein:DNA interaction on an EMSA. An antibody (typically 1 µg) is added to the binding reaction. During electrophoresis, the antibody:protein:DNA complex migrates slowly, causing a supershift compared to the shift caused by a protein:DNA complex. Not all antibodies will cause a supershift. Some antibodies do not bind to proteins once they are bound to DNA. Some antibodies can prevent protein:DNA interactions but can still be used to confirm the identity of a protein that causes a shift in the absence of the antibody.