ER-Tracker™ Red (BODIPY™ TR Glibenclamide), for live-cell imaging
Invitrogen™

ER-Tracker™ Red (BODIPY™ TR Glibenclamide), for live-cell imaging

ER-Tracker Red dye is cell-permeant, live-cell stain that is highly selective for the endoplasmic reticulum (ER) that when stained using深入閱讀
Have Questions?
產品號碼Quantity
E34250100 μg
產品號碼 E34250
價格 (HKD)
3,282.00
線上優惠
Ends: 31-Dec-2026
4,376.00
您節省 1,094.00 (25%)
Each
Quantity:
100 μg
價格 (HKD)
3,282.00
線上優惠
Ends: 31-Dec-2026
4,376.00
您節省 1,094.00 (25%)
Each
ER-Tracker Red dye is cell-permeant, live-cell stain that is highly selective for the endoplasmic reticulum (ER) that when stained using the protocol provided, the staining pattern is partially retained after fixation with formaldehyde. This stain consists of the green-fluorescent BODIPY TR dye and glibenclamide. Glibenclamide (glyburide) binds to the sulphonylurea receptors of ATP-sensitive K+ channels which are prominent on ER; the pharmacological activity of glibenclamide could potentially affect ER function. Variable expression of sulphonylurea receptors in some specialized cell types may result in non-ER labeling.

Visualize staining your cell without wasting your reagents, antibodies, or time with our new Stain-iT Cell Staining Simulator.

For Research Use Only. Not for use in diagnostic procedures.
規格
ColorRed
Detection MethodFluorescence
Excitation Wavelength Range587⁄615
For Use With (Equipment)Fluorescence Microscope
Product LineBODIPY, ER-Tracker
Quantity100 μg
Shipping ConditionRoom Temperature
Label TypeBODIPY Dyes
Product TypeDye
Sub Cellular LocalizationEndoplasmic Reticulum
Unit SizeEach
內容物與存放
Store in freezer -5°C to -30°C and protect from light.

常見問答集 (常見問題)

Why don't I see a significant change in signal for my live-cell fluorescent indicator dye?

Regardless of the type of live-cell indicator dye (e.g., calcium indicators, pH indicator, metal ion indicators), make sure there is no serum during the loading step, which can prematurely cleave dyes with AM esters and bind dyes non-specifically. Always optimize the dye concentration and staining time with a positive control before you run your test samples, to give the best signal-to-background. Always run a positive control with a buffer containing free ions of known concentration and an ionophore to open pores to those ions (for instance, for calcium indicators like Fluo-4 AM, this would include a buffer with added calcium combined with calcimycin, or for pH indicators, buffers of different pHs combined with nigericin). Reactive oxygen indicators, such as CellROX Green or H2DCFDA would require a cellular reactive oxygen species (ROS) stimulant as a positive control, such as menadione. Finally, make sure your imaging system has a sensitive detector. Plate readers, for instance, have much lower detector efficiency over background, compared to microscopy or flow cytometry.

Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.

引用資料與參考文獻 (28)

引用資料與參考文獻
Abstract
Binding of guanylyl cyclase activating protein 1 (GCAP1) to retinal guanylyl cyclase (RetGC1). The role of individual EF-hands.
Authors:Peshenko IV, Olshevskaya EV, Dizhoor AM,
Journal:J Biol Chem
PubMed ID:18541533
'Guanylyl cyclase activating protein 1 (GCAP1), after substitution of Ca(2+) by Mg(2+) in its EF-hands, stimulates photoreceptor guanylyl cyclase, RetGC1, in response to light. We inactivated metal binding in individual EF-hands of GCAP1 tagged with green fluorescent protein to assess their role in GCAP1 binding to RetGC1 in co-transfected HEK293 ... More
Imaging secretory vesicles by fluorescent protein insertion in propeptide rather than mature secreted peptide.
Authors:Watkins S, Geng X, Li L, Papworth G, Robbins PD, Drain P
Journal:Traffic
PubMed ID:12047554
'We combined confocal and live-cell imaging with a novel molecular strategy aimed at revealing mechanisms underlying glucose-regulated insulin vesicle secretion. The ''Ins-C-GFP'' reporter monitors secretory peptide targeting, trafficking, and exocytosis without directly tagging the mature secreted peptide. We trapped a green fluorescent protein (GFP) reporter in equimolar quantity within the ... More
Faecal excretion of ciprofloxacin after a single oral dose and its effect on faecal bacteria in healthy volunteers.
Authors:Pecquet S, Ravoire S, Andremont A,
Journal:J Antimicrob Chemother
PubMed ID:2211433
'High concentrations of ciprofloxacin have been shown to persist in the faeces of volunteers for several days after a week of oral treatment with this drug, which was also found to have a prolonged effect on aerobic Gram-negative intestinal bacteria. To determine whether a shorter course of ciprofloxacin would have ... More
Insulin-like growth factor I enhances the expression of aromatase P450 by inhibiting autophagy.
Authors:Zhang B, Shozu M, Okada M, Ishikawa H, Kasai T, Murakami K, Nomura K, Harada N, Inoue M,
Journal:Endocrinology
PubMed ID:20668023
'Aromatase, a key enzyme of estrogen biosynthesis, is transcriptionally regulated by many growth factors. IGF-I enhances aromatase activity in a variety of cells, but the mechanism of action has not been determined. We herein report our finding of a novel mechanism of action for IGF-I. IGF-I enhanced the dexamethasone (DEX)-induced ... More
Essential requirement for sphingosine kinase 2 in a sphingolipid apoptosis pathway activated by FTY720 analogues.
Authors:Don AS, Martinez-Lamenca C, Webb WR, Proia RL, Roberts E, Rosen H
Journal:J Biol Chem
PubMed ID:17400555
'The clinical immunosuppressant FTY720 is a sphingosine analogue that, once phosphorylated by sphingosine kinase 2 (Sphk2), is an agonist of multiple receptor subtypes for sphingosine 1-phosphate. Short exposures to FTY720 afford long term protection in lymphoproliferative and autoimmune disease models, presumably by inducing apoptosis in subsets of cells essential for ... More