Epidermal Growth Factor, Biotinylated, complexed to Alexa Fluor™ 555 Streptavidin (Alexa Fluor™ 555 EGF complex)
Invitrogen™

Epidermal Growth Factor, Biotinylated, complexed to Alexa Fluor™ 555 Streptavidin (Alexa Fluor™ 555 EGF complex)

We offer several conjugates of epidermal growth factor that are useful for the detection of EGF receptor in cells. Fluorescently자세히 알아보기
Have Questions?
카탈로그 번호수량
E35350
E-35350으로도 사용됨
100 μg
카탈로그 번호 E35350
E-35350으로도 사용됨
제품 가격(KRW)
684,000
온라인 행사
Ends: 30-Sep-2026
804,000
할인액 120,000 (15%)
Each
수량:
100 μg
제품 가격(KRW)
684,000
온라인 행사
Ends: 30-Sep-2026
804,000
할인액 120,000 (15%)
Each
We offer several conjugates of epidermal growth factor that are useful for the detection of EGF receptor in cells. Fluorescently labeled EGF has enabled scientists to investigate receptor-membrane interactions, study receptor distribution, calculate rate constants for the interaction of EGF with its receptor, and more. The conjugates incorporating fluorescein (FITC), Oregon Green™ 514, and tetramethylrhodamine were constructed with the dye directly attached to EGF. The conjugates of EGF with Alexa Fluor™ dyes and Texas Red™ dye are complexes of the dye–streptavidin molecule and biotinylated EGF. The biotinylated conjugates utilize biotin-XX, which contains a long spacer arm designed to enhance the probe’s affinity for the EGF receptor.

Epidermal Growth Factor Conjugate Specifications:
• EGF molecule: 53 amino acids, MW=6,045 Da
• Label (Ex/Em): Biotin-XX complexed with Alexa Fluor™ 555 streptavidin (∼555/565 nm)
• Number of fluorophore molecules on each EGF molecule: ∼2–3
• Fluorescence is typically monitored using a flow cytometer, fluorescence microscope, or fluorimeter


Find More Receptor Binding and Phagocytosis Probes
We offer a number of fluorescently labeled probes for studying receptor-mediated endocytosis, membrane markers for endocytosis and exocytosis, and methods for detecting internalized fluorescent ligands. Review Probes for Following Receptor Binding and Phagocytosis—Section 16.1 in the Molecular Probes™ Handbook for more information on these products.

For Research Use. Not for human or animal therapeutic or diagnostic use.
For Research Use Only. Not for use in diagnostic procedures.
사양
세포 유형Mammalian Cells
검출 방법Fluorescence
라벨 또는 염료Alexa Fluor Dyes
제품라인Alexa Fluor
수량100 μg
배송 조건Wet Ice
형태Lyophilized
제품 유형Growth Factor
Unit SizeEach
구성 및 보관
Store in freezer (-5 to -30°C) and protect from light.

인용 및 참조 문헌 (19)

인용 및 참조 문헌
Abstract
Regulation of endocytosis via the oxygen-sensing pathway.
Authors:Wang Y, Roche O, Yan MS, Finak G, Evans AJ, Metcalf JL, Hast BE, Hanna SC, Wondergem B, Furge KA, Irwin MS, Kim WY, Teh BT, Grinstein S, Park M, Marsden PA, Ohh M,
Journal:Nat Med
PubMed ID:19252501
'Tumor hypoxia is associated with disease progression, resistance to conventional cancer therapies and poor prognosis. Hypoxia, by largely unknown mechanisms, leads to deregulated accumulation of and signaling via receptor tyrosine kinases (RTKs) that are critical for driving oncogenesis. Here, we show that hypoxia or loss of von Hippel-Lindau protein--the principal ... More
Human ESCRT-II complex and its role in human immunodeficiency virus type 1 release.
Authors:Langelier C, von Schwedler UK, Fisher RD, De Domenico I, White PL, Hill CP, Kaplan J, Ward D, Sundquist WI
Journal:J Virol
PubMed ID:16973552
'The budding of many enveloped RNA viruses, including human immunodeficiency virus type 1 (HIV-1), requires some of the same cellular machinery as vesicle formation at the multivesicular body (MVB). In Saccharomyces cerevisiae, the ESCRT-II complex performs a central role in MVB protein sorting and vesicle formation, as it is recruited ... More
Two di-leucine motifs regulate trafficking of mucolipin-1 to lysosomes.
Authors:Vergarajauregui S, Puertollano R
Journal:Traffic
PubMed ID:16497227
'Mutations in the mucolipin-1 gene have been linked to mucolipidosis type IV, a lysosomal storage disorder characterized by severe neurological and ophthalmologic abnormalities. Mucolipin-1 is a membrane protein containing six putative transmembrane domains with both its N- and C-termini localized facing the cytosol. To gain information on the sorting motifs ... More
Rab14 is part of the early endosomal clathrin-coated TGN microdomain.
Authors:Proikas-Cezanne T, Gaugel A, Frickey T, Nordheim A,
Journal:FEBS Lett
PubMed ID:16962593
'Rab14 localizes to the Golgi/TGN and to early endosomes, but its biological function remains unclear. By structural modeling, we identified Rab14-specific residues and established a close relationship between the Rab2/Rab4/Rab14, Rab11/25 and Rab39 sub-groups within the Rab protein family. By quantitative confocal microscopy and by density centrifugation we show that ... More
Interactions of TOM1L1 with the multivesicular body sorting machinery.
Authors:Puertollano R
Journal:J Biol Chem
PubMed ID:15611048
'Tom1L1 (Tom1-like1) and related proteins Tom1 (Target of Myb1) and Tom1L2 (Tom1-like2) constitute a new protein family characterized by the presence of a VHS (Vps27p/Hrs/Stam) domain in the N-terminal portion followed by a GAT (GGA and Tom) domain. Recently it was demonstrated that the GAT domain of both Tom1 and ... More