Ethidium Homodimer-2 (EthD-2) - 1 mM Solution in DMSO
The cell-impermeant viability indicator ethdium homodimer-2 (EthD-2) is a high-affinity nucleic acid stain that is weakly fluorescent until bound to자세히 알아보기
Have Questions?
카탈로그 번호
수량
E3599
200 μL
카탈로그 번호 E3599
제품 가격(KRW)
554,000
온라인 행사
Ends: 30-Sep-2026
651,000
할인액 97,000 (15%)
Each
수량:
200 μL
제품 가격(KRW)
554,000
온라인 행사
Ends: 30-Sep-2026
651,000
할인액 97,000 (15%)
Each
The cell-impermeant viability indicator ethdium homodimer-2 (EthD-2) is a high-affinity nucleic acid stain that is weakly fluorescent until bound to DNA and emits red fluorescence (excitation/emission maxima ∼535/624).
Note: A protocol is not included with this classic reagent as they are readily available in the literature. The reagent is provided already dissolved at 1 mM in DMSO. A typical protocol for staining dead cells is to prepare a working solution at 0.1 to 10 μM in a physiological buffer and incubate with cells for 30-45 minutes at room temperature before analyzing on a fluorescent microscope, flow cytometer, or plate reader.
For Research Use Only. Not for use in diagnostic procedures.
사양
색상Orange
설명Ethidium Homodimer-2 (EthD-2) - 1 mM Solution in DMSO
검출 방법Fluorescence
염료 유형Cell-Impermeant
방출624 nm
여기 파장 범위535 nm
용도(애플리케이션)Cell staining assays
용도(장비)Fluorescence Microscope
수량200 μL
배송 조건Room Temperature
라벨 유형Fluorescent Dye
제품 유형Viability Indicator
Sub Cellular LocalizationNucleic Acids, Nucleus
Unit SizeEach
구성 및 보관
Store in freezer at -5°C to -30°C and protect from light.
인용 및 참조 문헌 (52)
인용 및 참조 문헌
Abstract
Differential effects of deuterium oxide on the fluorescence lifetimes and intensities of dyes with different modes of binding to DNA.
Authors:Sailer BL, Nastasi AJ, Valdez JG, Steinkamp JA, Crissman HA
Journal:J Histochem Cytochem
PubMed ID:9016307
'Deuterium oxide (D2O) increases both the fluorescence lifetime and the fluorescence intensity of the intercalating dyes propidium iodide (PI) and ethidium bromide (EB) when bound to nucleic acid structures. We have used spectroscopic analysis coupled with conventional and phase-sensitive flow cytometry to compare the alterations in intensity and lifetime of ... More
Diverse microglial motility behaviors during clearance of dead cells in hippocampal slices.
Authors:Petersen MA, Dailey ME
Journal:Glia
PubMed ID:15042586
'We used two-channel three-dimensional time-lapse fluorescence confocal imaging in live rat hippocampal slice cultures (1-7 days in vitro) to determine the motility behaviors of activated microglia as they engage dead and dying cells following traumatic brain tissue injury. Live microglia were labeled with a fluorescently conjugated lectin (IB(4)), and dead ... More
Calmodulin antagonists differentially affect capacitation-associated protein tyrosine phosphorylation of mouse sperm components.
Authors:Zeng HT, Tulsiani DR
Journal:J Cell Sci
PubMed ID:12668727
'Sperm capacitation in vitro is thought to be correlated with the increased protein tyrosine phosphorylation of a subset of sperm components. Our group recently used a pharmacological approach to demonstrate that calmodulin (CaM), a 17 kDa calcium sensor protein, has a role in sperm capacitation. In the present study, we ... More
Automated image analysis of live/dead staining of the fungus Aureobasidium pullulans on microscope slides and leaf surfaces.
Authors:Nelson CD, Spear RN, Andrews JH
Journal:Biotechniques
PubMed ID:11056819
'An image analysis program and protocol for the identification and enumeration of live versus dead cells of the yeast-like fungus Aureobasidium pullulans was developed for both populations on microscope slides and leaf surfaces. Live cells took up CellTracker Blue, while nonviable cells stained with DEAD Red. Image analysis macro programs ... More
A2E-epoxides damage DNA in retinal pigment epithelial cells. Vitamin E and other antioxidants inhibit A2E-epoxide formation.
Authors:Sparrow JR, Vollmer-Snarr HR, Zhou J, Jang YP, Jockusch S, Itagaki Y, Nakanishi K
Journal:J Biol Chem
PubMed ID:12646558
'The autofluorescent pigments that accumulate in retinal pigment epithelial cells with aging and in some retinal disorders have been implicated in the etiology of macular degeneration. The major constituent is the fluorophore A2E, a pyridinium bisretinoid. Light-exposed A2E-laden retinal pigment epithelium exhibits a propensity for apoptosis with light in the ... More