Thermo Scientific™
RA Lamb High-Quality Plastic Slide Boxes
Related applications:
Anatomical Pathology
Store and protect up to 100 microscope slides with Thermo Scientific™ RA Lamb High-Quality Plastic Slide Boxes.
Back to top
  • Boxes are grooved to snugly store either 25 or 100 76 x 26mm slides, and a close-fitting snap-on lid keeps everything secure
  • Impact-resistant polystyrene construction protects your most important slides
  • Separate card index organizes your slides
Please Note: Availability may vary by country.