Thermo Scientific™
ColorStack Storage System
Related applications:
Anatomical Pathology
Achieve higher levels of organization when archiving slides with Thermo Scientific™ ColorStack Storage Systems. An effective visual method of sorting, these systems are a perfect compliment to the standard cabinets, and can be used with the LabStack cabinet series. The cabinet is cream epoxy-coated metal. The color-coded drawers have colored plastic fronts; available in 10 different shades.
Back to top
Includes
  • 14 drawers of one color
  • Each of these storage drawers is 48 x 48 x 14cm and can hold 6,000 slides.
Compatablity
  • Fully compatible with all standard cabinets, bases, and options
  • Colorstack drawers can easily replace drawers in a standard 14-drawer cabinet
  • Special top cover is cream to match the ColorStack 14-Drawer Cabinet