Thermo Scientific™
RA Lamb Mini Heavy-Duty Wheel Base Stand for LabStack Storage System Mini Cabinets
 Related applications:
Anatomical Pathology
Employ the same flexibility and customization as the standard LabStack range, but as a benchtop system. A more compact version of this versatile lab storage system, Thermo Scientific™ RA Lamb Mini Cabinets for LabStack Storage System and options are half the depth of standard cabinets to fit comfortably on a benchtop or in a small space.
Error loading your content!
  Catalog number Price ({{currency}}) Availability Qty
{{product.sku}}
also known as {{product.formattedSku}} 
{{product.availability.message}} Add To Cart
{{addItemToList.successMessage}} {{addItemToList.listName}}
Back to top
Description
Features:
  • Custom stacked storage with single cabinets that fit together securely
  • Easy removal, relocation, and removal of drawers
  • Dust-free storage ensured with epoxy-coated steel, sealed from top to bottom
Safety:
  • Stops prevent each drawer from accidentally sliding all the way out
  • Metal rim runs full width of both front and rear to lock securely with the next cabinet in the stack
Options:
  • Optional wheeled base unit raises the stack and provides mobility
  • Optional panel creates additional work space by adding a cover to the top of the stack
  • Several cabinet configurations available
Documents
Manuals & protocols