Thermo Scientific™
RA Lamb Horizontal Slide Filing Cabinets
Related applications:
Anatomical Pathology
Store up to 1230 76 x 26mm slides with Thermo Scientific™ RA Lamb Horizontal Filing Cabinets.
Back to top
  • Two drawers with grooved polypropylene dividers hold slides snugly in place
  • Ideal for large-volume laboratory work
  • Available in three colors to color-code large batches of slides
Please Note: Availability may vary by country.