Thermo Scientific™
RA Lamb Glass Coplin Jar, holds 5 slides (10 back to back)
 Related applications:
Anatomical Pathology
Hold up to 10 slides back-to-back with Thermo Scientific™ RA Lamb molded glass coplin jars. Jars have a drop-on lid and integral large diameter base.
Error loading your content!
  Catalog number Price ({{currency}}) Availability Qty
{{product.sku}}
also known as {{product.formattedSku}} 
{{product.availability.message}} Add To Cart
{{addItemToList.successMessage}} {{addItemToList.listName}}
Back to top
Description
Features
  • Heavy, clear glass construction with internal grooves
  • 1.6in. diameter circular tapered top
Includes
  • Plastic screw-on cap.
Documents
Manuals & protocols