Selenite Lactose Broth
Actual product may vary
Thermo Scientific™
Selenite Lactose Broth
Reduce preparation time when enriching for Salmonella species from fecal and food samples with ready-to-use prepared media tubes.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantityMedia TypeFill Volume
EB0213E100/Pk.Selenite F Broth100 mL
TV5005I50/Pk.Selenite Lactose Broth8 mL
Catalog number EB0213E
Price (EUR)
-
Quantity:
100/Pk.
Media Type:
Selenite F Broth
Fill Volume:
100 mL
Enrich for Salmonella species from fecal and food samples with Thermo Scientific™ Selenite Lactose Broth in convenient prepared media tubes.
Selenite Lactose Broth is used for enrichment culture of salmonellae from feces, food products, water and other materials.
Sodium biselenite acts as the selective agent, while lactose is added as a fermentable carbohydrate to prevent a rise in pH value during incubation because any increase in pH will reduce the selective activity of selenite.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.
For In Vitro Diagnostic Use.
Specifications
ColorLight ivory, transparent
Fill Volume100 mL
Fill Weight10 ± 0.5mL
FormatUniversal Tube
Media TypeSelenite F Broth
Packaging Type100 universals in a carton box
Quantity100/Pk.
Shelf Life26 Weeks
Manufacturing OriginScotland
Product TypeMicrobiology Media
Unit SizeEach
Contents & Storage
2°C to 10°C