Bolton Broth
Bolton Broth
Thermo Scientific™

Bolton Broth

Selectively enrich Campylobacter species (such as C. jejuni and C. coli) from food and environmental samples prior to isolation on selective agar.
Have Questions?
Catalog NumberFill Weight
EB1070W1000 to 1005 g
Catalog number EB1070W
Price (MXN)
-
Request A Quote
Fill Weight:
1000 to 1005 g
Bolton Broth is used to selectively enrich Campylobacter species (such as C. jejuni and C. coli) from food and environmental samples prior to isolation on selective agar.
Specifications
ColorDark Red/Brown
Fill Weight1000 to 1005 g
Format1 L Sirop Bottle
Packaging Type4 Sirop Bottles with screw cap closures in a box
Quantity4/Pk.
Shelf Life13 Weeks
Manufacturing OriginUnited Kingdom
Product TypeMicrobiology Media
Unit SizeEach
Contents & Storage
2°C to 8°C