InhibiSURE™ Viral Inactivation Medium
InhibiSURE™ Viral Inactivation Medium
Thermo Scientific™
InhibiSURE™ Viral Inactivation Medium
Non-hazardous, inactivation media for viral collection and transport.
Have Questions?
Cambiar vistabuttonViewtableView
Número de catálogoTipo de producto
EB1372AMedio de inactivación vírica
SK01372Kit de hisopo NPG con medio de inactivación
Número de catálogo EB1372A
Precio (MXN)
-
Solicite un presupuesto
Tipo de producto:
Medio de inactivación vírica
Rapidly inactivate and stabilize viral RNA at ambient temperature with a non-hazardous formulation using Thermo Scientific™ InhibiSURE™ Viral Inactivation Medium. This liquid medium is intended for the collection, transportation, and inactivation of SARS-CoV-2 and RNA enveloped viruses.
Collect and inactivate RNA enveloped viruses with Thermo Scientific™ InhibiSURE™ Viral Inactivation Medium. InhibiSURE stabilises viral RNA for transportation and use in subsequent in vitro diagnostic testing procedures.  The device is intended to be used with unprocessed nasal swabs, nasopharyngeal swabs and throat swabs.  This is a single use device, it is not automated nor is it a companion diagnostic.
Product is CE marked but not 510(k)-cleared and not available for sale in the U.S. Availability of product in each country depends on local regulatory marketing authorization status. - For In Vitro Diagnostic Use.
Para uso diagnóstico in vitro
Especificaciones
Color del tapónPurple
CompatibilidadInhibiSURE™ Viral Inactivation Medium has been designed to be compatible with nucleic acid purification and amplification workflows. It has been validated with the Thermo Scientific™ MagMax™ MVP II kit (product code A48383) and TaqPath™ COVID 19 CE IVD RT PCR Kit (A48067)
DescripciónInhibiSURE™ Viral Inactivation Medium (1.5mL)
Volumen de llenado1.5 mL
Para utilizar con (aplicación)Medium is a liquid medium intended to be used for the collection and inactivation of RNA enveloped viruses. The product stabilises the viral RNA for transportation and use in subsequent in vitro diagnostic testing procedures. The device is intended to be used with unprocessed nasal swabs, nasopharyngeal swabs and throat swabs.
Cantidad100
Requisitos de almacenamiento 2 - 30°C
Temperatura de transporte2 - 30°C
Tipo de productoMedio de inactivación vírica
Unit SizeEach