Search
| 產品號碼 | Length (Metric) Cable |
|---|---|
| EC620102 | 1 m |
Eutech™ Conductivity/Temperature Electrode EC620102 is used in conductivity meters in various applications. Get quick and accurate conductivity measurements in the field, laboratories, and schools.
Features