Product Image
Thermo Scientific™

pH Buffer Sachets

Get a wide range of Eutech™ pH buffers and solutions for use with pH meters and electrodes
Have Questions?
Change viewbuttonViewtableView
Catalog NumberChemical Name or MaterialpH
ECBU10BSpH Buffer Sachet10.01
ECRINWTpH Deionized Water RinseNot Applicable
Catalog number ECBU10BS
Price (USD)
71.40
Each
Chemical Name or Material:
pH Buffer Sachet
pH:
10.01
Price (USD)
71.40
Each

pH Buffer Sachets are NIST-traceable sachets for quick, accurate and convenient calibrations. Simply insert electrode into sachet pack, calibrate, rinse and discard sachet – no separate container needed.

Features:

  • pH Buffer Sachets -20 ml
  • NIST Traceable
Specifications
Certifications/ComplianceNIST Traceable
Chemical Name or MaterialpH Buffer Sachet
Quantity20 x 20 mL
Solution TypeBuffer
pH10.01
Unit SizeEach