Eutech™ pH Buffer Tablets
Actual product may vary
Eutech™ pH Buffer Tablets
Eutech™ pH Buffer Tablets
Thermo Scientific™
Eutech™ pH Buffer Tablets
Eutech™ offers pH buffer tablets for use with pH meters and electrodes
Have Questions?
보기 방식 변경buttonViewtableView
카탈로그 번호pH
ECBU7BX7
ECBU4BX4
ECBU10BX10
카탈로그 번호 ECBU7BX
제품 가격(KRW)
83,000
Each
pH:
7
제품 가격(KRW)
83,000
Each
Eutech™ pH buffer tablets provide a quick, convenient way to prepare pH buffers from tablets for pH calibrations. Each box contains 100 pH buffer tablets.
Eutech pH buffer tablets are designed to provide a quick and simple way to prepare pH buffers from tablets. Mix 30 ml of deionized or distilled water with each tablet. When prepared correctly, the resulting pH buffer solution will have a tolerance of +/- 0.05 pH.
  • Tablets are individually packaged for long shelf life storage
  • Tablets are easy to prepare
  • Tablets are lightweight and compact, ideal for field use
사양
색상Yellow
용도(애플리케이션)pH calibration
용도 (장비)pH Meter and Electrode
물리적 형태Tablet
수량100 Tablets
용액 유형Buffer
pH7
Unit SizeEach