Eutech™ Conductivity Electrodes are used in conductivity meters in various applications. Get quick and accurate conductivity measurements in the field, laboratories, and schools.
Features:
- Parameter: Conductivity/ATC cell
- material: Glass
- Cell Constant: K = 0.1
- Cable Length: 1m
- Connector: DIN connector
- Cable Length: 1m
- Used With: CyberScan CON 510, Eutech CON 150